Skip to Content
MilliporeSigma

Skip To

GF00524607

Tetrafluoroethylene-Perfluoro(alkoxy vinyl ether)

(alkoxy vinyl ether), (pfa), film, thickness 0.025 mm, L 5 m

Synonym(s):

FV321025

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
$854.00

About This Item

NACRES:
NA.23
UNSPSC Code:
12352100
Manufacturer/tradename:
Goodfellow 005-246-07

$854.00


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

film

manufacturer/tradename

Goodfellow 005-246-07

L

5 m

thickness

0.025 mm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF57301769GF38897263GF52374618
description

(alkoxy vinyl ether), (pfa), film, thickness 0.025 mm, L 5 m

description

(alkoxy vinyl ether), (pfa), film, thickness 0.125 mm, size 100 × 100 mm, natural

description

(alkoxy vinyl ether), (pfa), film, thickness 0.125 mm, size 200 × 200 mm, natural

description

(alkoxy vinyl ether), (pfa), film, thickness 0.075 mm, size 400 × 400 mm, natural

manufacturer/tradename

Goodfellow 005-246-07

manufacturer/tradename

Goodfellow 573-017-69

manufacturer/tradename

Goodfellow 388-972-63

manufacturer/tradename

Goodfellow 523-746-18

thickness

0.025 mm

thickness

0.125 mm

thickness

0.125 mm

thickness

0.075 mm

L

5 m

L

-

L

-

L

-

form

film

form

film

form

film

form

film


Still not finding the right product?


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters