Skip to Content
MilliporeSigma

GF44249698

Tungsten-rhenium alloy, W75Re25

0.05m wire, 0.75mm diameter, annealed

Synonym(s):

W 015145

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
11101711
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

wire

composition

W75Re25

reaction suitability

core: rhenium, core: tungsten

manufacturer/tradename

Goodfellow 442-496-98

L × diam.

0.05 m × 0.75 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow


Still not finding the right product?

Explore all of our products under Tungsten-rhenium alloy, W75Re25




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library