Skip to Content
MilliporeSigma

GF87397015

Aluminum-cobalt alloy, Al99Co1

1000mm wire, 0.5mm diameter

Synonym(s):

AB235120

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352311
eCl@ss:
38120507
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

wire

composition

Al99Co1

reaction suitability

core: aluminum, core: cobalt

manufacturer/tradename

Goodfellow 873-970-15

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow


Still not finding the right product?

Explore all of our products under Aluminum-cobalt alloy, Al99Co1




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library