Skip to Content
MilliporeSigma

Skip To

GF92597451

Copper

foil, thickness 0.00075 mm, size 25 × 25 mm, purity 99.97%

Synonym(s):

CU000090

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
$2,010.00
5 ea
Please contact Customer Service for Availability
$8,140.00

About This Item

Linear Formula:
Cu
CAS Number:
Molecular Weight:
63.55
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141711
MDL number:
eCl@ss:
38150101

$2,010.00


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99.97%

form

foil

manufacturer/tradename

Goodfellow 925-974-51

resistivity

1.673 μΩ-cm, 20°C

size

25 × 25 mm

thickness

0.00075 mm

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF51053699GF86241719GF94916188
description

foil, thickness 0.00075 mm, size 25 × 25 mm, purity 99.97%

description

tube, 100mm, outside diameter 3.0mm, inside diameter 1.2mm, wall thickness 0.9mm, as drawn, 99.9%

description

foil, thickness 0.0015 mm, purity 99.97%, size 25 × 25 mm

description

foil, thickness 0.05 mm, length 50 m, coil width 12 mm

form

foil

form

tubes

form

foil

form

foil

assay

99.97%

assay

99.9%

assay

99.97%

assay

-

manufacturer/tradename

Goodfellow 925-974-51

manufacturer/tradename

Goodfellow 510-536-99

manufacturer/tradename

Goodfellow 862-417-19

manufacturer/tradename

Goodfellow 949-161-88

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C

resistivity

1.673 μΩ-cm, 20°C


Storage Class

13 - Non Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters