Skip to Content
MilliporeSigma

BI0233

Capping Reagent A 5% Phenoxyacetic anhydride w/v in Tetrahydrofuran with 10% Pyridine v/v

Novabiochem®

Synonym(s):

THF/Pyr/Pac2O (85:10:5)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116006
eCl@ss:
32035490
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Novabiochem®

form

liquid

manufacturer/tradename

Novabiochem®

color

yellow to yellow

density

0.914 g/mL

shipped in

ambient

storage temp.

room temp

General description

Biosynthesis Novabiochem®

Application

Chemical synthesis (Intermediate use)

Packaging

Glass bottle (Pkg Module 7.60003.4477)

Other Notes

Flash point: -13 °C

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

Flash Point (°F)

8.6 °F

Flash Point (°C)

-13 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library