Skip to Content
MilliporeSigma

SHF0650425

Hi-Flow™ Plus Membrane 65

4 mil backing, 25 mm width, pack of 1 roll

Synonym(s):

Hi-Flow, HF, lateral flow membrane, immunoassay membrane, nitrocellulose membrane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32031602
UNSPSC Code:
40161507
NACRES:
NA.82
Packaging:
pack of 1 roll
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

packaging

pack of 1 roll

parameter

65 sec/4 cm capillary flow rate

L

100 m (328 ft)

thickness

4 mil (100 μm)

width

2.5 cm

General description

Hi-Flow™ Plus membranes are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. Lateral flow tests faciliate quick and convenient detection / quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine. Hi-Flow™ Plus membranes are ideally suited for such applications. Available in a variety of capillary flow times and backing options, choosing Hi-Flow™ Plus membranes can improve assay performance and offer flexibility to meet a wide range of assay design requirements. Hi-Flow™ Plus 65 (HF065) is suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern. This Hi-Flow™ Plus 65 pack contains one 25mm x 100m slit roll of 4 mil backed nitrocellulose membrane with a nominal capillary flow time of 65 seconds/4 cm.

Application

  • Hi-Flow™ Plus membrane is a critical raw material suitable for use in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women′s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing and animal health.
  • Hi-Flow™ Plus membranes are suitable for use with all lateral flow test techniques including competitive, sandwich, and multiplex detection assays.
  • They are optimised for use in both qualitative and quantitative test types and pair well with lateral flow detector particles including colloidal gold nanoparticles, dyed/ fluorescent microspheres etc.
  • Hi-Flow™ Plus membranes are suitable for use in lateral flow tests designed to detect analytes present in a broad range of samples matrices including, blood, saliva, urine, nasopharyngeal swabs, sweat and others.

Features and Benefits

FEATURES:

  • Consistent Flow & Surface Chemistry, Robust manufacturing process
  • Proprietary Surface Chemistry Optimised for Lateral Flow Applications
  • High Quality Level: MQ400
  • Trusted Partner and Brand with Decades of membrane manufacturing experience
  • High production capacity, high stock levels, multiple manufacturing lines
  • Convenient samples packs available in several formats
  • Manufacturing scale quantities available to purchase online with immediate shipping.
  • We offer a complete solution; along with Hi-Flow Plus membranes we offer raw materials across the entire lateral flow workflow as well as assay development services (include link to Lateral Flow Assay Development Services (sigmaaldrich.com).



BENEFITS:

  • Consistent capillary flow time and membrane thickness
  • Superior surface morphology allows for consistent and even flow distribution of reagents across the membrane surface
  • Sharp, intense, and uniform test and control line formation
  • Excellent intra-lot and inter-lot consistency
  • Better Assay sensitivity
  • Excellent reproducibility
  • Improved lateral flow test manufacturing yields
  • Quick Delivery and Lead time
  • No supply disruption
  • Excellent Technical support

Legal Information

Hi-Flow is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under HI-Flow™ Plus Membrane 65


Signal Word

Danger

Hazard Codes

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library