Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.24
Pricing and availability is not currently available.
grade
analytical standard
description
Seraseq® Reference Material
manufacturer/tradename
LGC
application(s)
clinical testing
storage temp.
−20°C
General description
A cfDNA extraction reference material at a defined concentration of 80 ng/mL, used to standardize and validate cfDNA extraction workflows for liquid biopsy assays.
This product is provided as delivered and specified by the manufacturer. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued by the manufacturer. For further information and support, please go to the website of the manufacturer at https://www.seracare.com/Controls-Reference-Materials/
Analysis Note
These products are for research use only and not for use in diagnostic procedures.
Other Notes
Sales restrictions may apply.
Legal Information
Seraseq is a registered trademark of LGC Clinical Diagnostics, Inc.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation
Target Organs
Respiratory Tract
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

