Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
UNSPSC Code:
12352104
MDL number:
Beilstein/REAXYS Number:
635639
NACRES:
NA.21
Assay:
≥99.5%
Grade:
anhydrous
Bp:
82 °C (lit.), 82 °C
Vapor pressure:
33 mmHg ( 20 °C), 44 mmHg ( 25 °C)
grade
anhydrous
Quality Segment
vapor density
2.1 (vs air)
vapor pressure
33 mmHg ( 20 °C), 44 mmHg ( 25 °C)
assay
≥99.5%
form
liquid
autoignition temp.
750 °C, 750 °F
expl. lim.
2.0-12.7 %, 93 °C
sustainability
Sustainability features
renewable carbon content
78%
refractive index
n
pH
7 (20 °C)
bp
82 °C (lit.), 82 °C
mp
-89.5 °C, −89.5 °C (lit.)
solubility
soluble (water soluble completely)
density
0.785 g/mL at 25 °C (lit.), 0.785 g/mL at 25 °C
SMILES string
OC(C)C, CC(C)O
InChI
1S/C3H8O/c1-3(2)4/h3-4H,1-2H3
InChI key
KFZMGEQAYNKOFK-UHFFFAOYSA-N
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is biorenewable and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". Click here for more information. This anhydrous grade 2-propanol is a drop-in replacement for any 2-propanol application. Made from 78.0% renewable carbon (verified through C14 ASTM D6866-16 testing) and produced in an environmentally friendly way that reduces the carbon footprint of production and reliance on fossil fuels. Because it is not from petroleum, it is free from many non-renewable contaminants, such as benzene, phthalates, and other phenols, leading to a higher quality product.
Features and Benefits
Renewable Carbon Content >78.0% ASTM D6866 - Standard Test Methods for Determining the Biobased Content Made from Renewable Resource - corn waste Replacement for Petroleum-based 2-Propanol
Still not finding the right product?
Explore all of our products under 2-Propanol
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class
3 - Flammable liquids
WGK
WGK 1
Flash Point (°F)
53.6 °F - closed cup
Flash Point (°C)
12.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

