Skip to Content
MilliporeSigma

PMC1101

Ethyl alcohol, Pure

≥99.5%, ISOM8™, anhydrous, 200 (proof)

Sustainability features

Synonym(s):

Ethyl alcohol, Pure, Ethanol, absolute alcohol, non-denatured ethanol

Select a Size

Change View

About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
UNSPSC Code:
15101802
MDL number:
eCl@ss:
32039090
Assay:
(GC), ≥99.5%
Grade:
Elevated Assurance, anhydrous
Bp:
78.3 °C
Vapor pressure:
44 mmHg ( 20 °C)
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

Elevated Assurance, anhydrous

vapor density

1.59 (vs air)

vapor pressure

44 mmHg ( 20 °C)

product line

ISOM8™

assay

(GC), ≥99.5%

form

liquid

autoignition temp.

365 °C (689°F)

shelf life

36 mo.

expl. lim.

3.1-27.7 %

packaging

Sure/Seal™ of 1 L, Sure/Seal™ of 6 × 1 L, Sure/Seal™ of 100 mL, Sure/Seal™ of 4 × 2 L, pkg of ( Various Returnable Container Options!)

origin

origin

sustainability

Sustainability features

concentration

200 (proof)

impurities

<0.005% water

evapn. residue

<0.0005%

refractive index

n20/D 7 (10 g/L)

bp

78.3 °C

mp

-114 °C

transition temp

flash point 13 °C (55°F)

solubility

miscible

density

0.789 g/mL at 20 °C

cation traces

(See Dossier)

λ

(See Dossier)

application(s)

diagnostic assay manufacturing
life science and biopharma

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

General description

Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8 team, we have expertise and documentation to support ISO 9001 industrial and non-GMP pharma production needs, as defined by our M-Clarity™ Program.

Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich customers′ requirements. Please navigate to our documents section to look at dossiers and other quality documentation for this product, provided with support of our Elevated Assurance Program.

Our ISOM8™ Ethyl Alcohol is available within the USA with our ® Solvent System for returnable, reusable, and readily available bulk solvent delivery, to reduce packaging waste and help you achieve your sustainablility goals. To request ISOM8 and other product lines in returnable containers Click HereUnlock your manufacturing potential with our comprehensive program.

By choosing ISOM8™ chemicals, ensure compliance while fostering innovation and excellence. Connect with us to learn more about how we can support your journey toward manufacturing success.

Application

  • Pharmaceutical Development
  • Our ethanol serves as a versatile solvent, reagent, and intermediate in non-GMP pharmacuetical production.
  • Industrial Applications
  • Our pure ethanol is also employed in various industrial processes, including cleaning and degreasing, where a high-purity solvent is required.

Features and Benefits

FEATURES
  • Renewable Sourcing
  • Our anhydrous ethanol is derived from renewable feedstocks, reducing reliance on fossil fuels and promoting a circular economy.
  • High Purity
  • We ensure that our anhydrous ethanol meets stringent quality standards, making it suitable for a wide range of applications.
  • Versatile Applications
  • This product can be utilized across various industries, for production and purification.
  • Returnable Containers
  • Our anhydrous ethanol is offered in returnable containers, designed for multiple uses, which minimizes waste and supports sustainability practices.
BENEFITS
  • Waste Reduction
  • Utilizing returnable containers significantly reduces packaging waste, aligning with sustainability initiatives.
  • Cost Efficiency
  • Returnable containers can lead to cost savings over time, as customers can reuse them rather than purchasing new packaging for each order.
  • Regulatory Compliance
  • Our bio-based anhydrous ethanol aligns with global regulations, providing peace of mind.

Packaging

100 mL, 1 L, and 2.5 L bottles are packaged in Sure/Seal™ containers.
For bulk amounts in US, we offer returnables through ReCycler® Program

Legal Information

ISOM8™ is a trademark of Sigma-Aldrich Co. LLC
ISOM8 is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany
RECYCLER is a registered trademark of Merck KGaA, Darmstadt, Germany
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?

Explore all of our products under Ethyl alcohol, Pure


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

Flash Point (°F)

55.4 °F - closed cup

Flash Point (°C)

13 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library