Skip to Content

CM3.B6

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
human blood
Growth mode:
Suspension
Karyotype:
Not specified
Morphology:
Small rounded cells
Manufacturer/tradename:
ECACC
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human blood

manufacturer/tradename

ECACC

packaging

tube of 5 μg 11060101-DNA-5UG, pkg of vial of cells 11060101-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Small rounded cells

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

-140 to -196°C

Biochem/physiol Actions

Human B-lymphocyte cell line, immortalized with Epstein-Barr Virus (EBV), derived from a patient with chronic Hepatitis C (HCV type 1). Cell line produces human monoclonal antibody specific for non-structural protein NS3 (helicase domain). This antibody binds to a major conformational epitiope located at the carboxyl-terminal end of the NS3 region that is recognised by approximately 90% from HCV-infected patients. CM3.B6 can be used in immunochemistry, HCV protein purification and HCV typing. CM3.B6 and B12.F8 (Sigma product number 11060102) were derived from the same patient as confirmed by STR profiling.
Human anti-hepatitis C virus IgG1 secreting B cell
STR-PCR Data:

Amelogenin: X
CSF1PO: 11,12
D13S317: 9,11
D16S539: 11,12
D5S818: 11,13
D7S820: 8,11
THO1: 6
TPOX: 9,11
vWA: 14,16

Preparation Note

RPMI 1640 (R0883) + 2 mM L-L-Glutamine (G7513) + 1% NEAA (M7145) + 1% NaP (S8636) + 10% FBS / FCS (F2442) + 20U/ml Interlukin-6 (IL-6).
Maintain cultures between 3-9 x 105/ml. 5% CO2; 37 °C. Media change every two days. Saturation density 1.1 x 106/ml

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). Use of this product by any organization may require a license agreement. Customers will be contacted by cellscreening@merckgroup.com to provide further information after the order request is submitted. Any other use of these products is prohibited without the express written permission of UKHSA.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). Use of this product by commercial organizations may require a license agreement. Commercial customers will be contacted by cellscreening@merckgroup.com to provide further information after the order request is submitted. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library