Skip to Content
MilliporeSigma

YTA 74.4.4

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 11060804

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
Rat x Rat Hybridoma
Isotype:
IgG2a
Technique(s):
cell culture | mammalian: suitable
Myeloma:
Y3.AG.1.2.3
Reactivity:
Mouse transferrin receptor (CD71)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Rat x Rat Hybridoma

manufacturer/tradename

ECACC

packaging

tube of 5 μg 11060804-DNA-5UG, pkg of vial of cells 11060804-1VL

ImmunoDon

DA rat spleen

Myeloma

Y3.AG.1.2.3

Reactivity

Mouse transferrin receptor (CD71)

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

-140 to -196°C

Immunogen

Activated CBA mouse splenic T cells

Biochem/physiol Actions

Monoclonal antibody human CD25
Rat hybridoma producing antibody against mouse CD7. Cambridge Enterprise Reference number AB110:www.enterprise.cam.ac.uk.

Preparation Note

IDMEM (I3390) + 2 mM L-Glutamine (G7513) + 5% FBS / FCS (F2442). Where the cells are grown under static conditions, 0.02% EDTA, Trypsin (T4424) or mechanical agitation maybe required to dislodge them from the plastic.
Maintain cultures between 3-9 x 105 cells/ml. 5% CO2; 37 °C. Saturation density 9 x 105/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). Use of this product by commercial organizations may require a license agreement. Commercial customers will be contacted by cellscreening@merckgroup.com to provide further information after the order request is submitted. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library