Skip to Content
MilliporeSigma

EP0030113632

Eppendorf® Twin tec PCR plates, 96 wells

skirted 2D SafeCode, colorless wells, red Plate, pack of 25 plates (5 bags × 5 plates)

Synonym(s):

culture plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106300
eCl@ss:
32039090
Sterility:
non-sterile
Feature:
PCR clean, skirt
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

feature

PCR clean, skirt

packaging

pack of 25 plates (5 bags × 5 plates)

manufacturer/tradename

Eppendorf® 0030113632

well maximum volume

150 μm

well size

96 wells

color

colorless wells, red Plate

General description

Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.

Legal Information

Eppendorf is a registered trademark of Eppendorf SE


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library