Skip to Content
MilliporeSigma

EP3102663005

Eppendorf® Research 3 neo ColorTag

34mm, light violet, 5 pieces

Synonym(s):

label, color identification bands, pipette identification rings

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 5 pieces

manufacturer/tradename

Eppendorf® 3102663005

W

34 mm

color

violet, light

compatibility

Eppendorf® pipettes

Legal Information

Eppendorf is a registered trademark of Eppendorf SE


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library