Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
Please contact Customer Service for Availability | $651.00 |
About This Item
UNSPSC Code:
41116104
eCl@ss:
40050204
$651.00
Please contact Customer Service for Availability
usage
sufficient for 400 fluorometric tests
detection method
fluorometric
storage temp.
−20°C
General description
Farnesyltransferase (FTase, EC 2.5.1.58) catalyzes the transfer of a farnesyl group from farnesylpyrophosphate to the cysteine residue of the C-terminus of target proteins. When not properly regulated, farnesylated proteins, including the Ras super family of small GTPases, can lead to developmental disorders and cancer.
Application
Suitable for the quantitative determination of FTase enzyme activity in biological samples.
Biochem/physiol Actions
In this assay, FTase reacts with farnesyl pyrophosphate and the dansyl-peptide substrate releasing a product that is measurable by fluorescence (λem/ex = 340/550 nm). The linear detection range is 0.024 – 3.2 U/LFTase.
1 of 1
This Item | |||
|---|---|---|---|
| description sufficient for 400 fluorometric tests | description sufficient for 400 fluorometric tests | description Supplied by Roar Biomedical, Inc., sufficient for 100 fluorometric tests | description Supplied by Roar Biomedical, Inc. |
| usage sufficient for 400 fluorometric tests | usage sufficient for 400 fluorometric tests | usage sufficient for 100 fluorometric tests | usage sufficient for 100 fluorometric tests |
| detection method fluorometric | detection method fluorometric | detection method fluorometric | detection method fluorometric |
| storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
Storage Class
10 - Combustible liquids
WGK
nwg
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
