Skip to Content
MilliporeSigma

MIR22

TransIT®-mRNA Transfection Kit

A high efficiency, low toxicity transfection reagent for large RNA and CRISPR guide RNA

Synonym(s):

MIR 22, mRNA transfection

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106502
Form:
liquid
Grade:
Molecular Biology
Technique(s):
transfection: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

TransIT®-mRNA, A high efficiency, low toxicity transfection reagent for large RNA and CRISPR guide RNA

grade

Molecular Biology

form

liquid

manufacturer/tradename

Mirus Bio

technique(s)

transfection: suitable

suitability

suitable for CRISPR gene editing

shipped in

wet ice

storage temp.

2-8°C

General description

TransIT®-mRNA Transfection Kit is designed to transfect RNA into a broad range of cell types with minimal cellular toxicity. RNA delivery avoids transcriptional regulation effects by directly delivering the RNA to the cytoplasm for expression. The TransIT-mRNA Transfection Kit can be used to deliver a variety of RNA molecules including mRNAs and viral RNAs (2–10 kb). It can be used for multiple applications such as short-term protein expression, viral production and replication studies. TransIT®-mRNA Transfection Kit contains two components, namely: TransIT®-mRNA Reagent and the mRNA Boost Reagent. This kit is compatible with serum; transfection efficiency is optimal when transfections are performed in the presence of serum, with no medium change required.

Features and Benefits

Each Kit is supplied with the TransIT®-mRNA Transfection Reagent and the mRNA Boost Reagent
  • Low Cellular Toxicity – Maintain cell density and reduce experimental biase
  • High Efficiency Delivery – Achieve RNA delivery in a large population of cells to ensure experimental success
  • Serum Compatible – Perform transfections in the presence of serum which eliminates the need for a media change and maintains cellular health
  • Deliver Various Sizes of RNA – Ideal for specialized applications, such as virus production, protein expression and CRISPR genome editing

Other Notes

Biological Source: raw materials that are derived from animals.

Suitable for:
  • CRISPR/Cas genome editing
  • Stem cell transfection
  • VRNA transfection
  • Co-transfection

Legal Information

TransIT is a registered trademark of Mirus Bio




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library