Skip to Content
MilliporeSigma

MIR540

TransIT®-2020 Transfection Reagent

A high performance, animal-free, broad spectrum DNA transfection reagent

Synonym(s):

DNA transfection, MIR 540

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106502
Form:
liquid
Grade:
Molecular Biology
Technique(s):
transfection: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

TransIT®-2020, A high performance, animal-free, broad spectrum DNA transfection reagent

grade

Molecular Biology

form

liquid

manufacturer/tradename

Mirus Bio

technique(s)

transfection: suitable

shipped in

wet ice

storage temp.

−20°C

General description

TransIT®-2020 Transfection Reagent is a high-performance, animal-origin free, broad spectrum reagent that provides exceptional transfection of plasmid DNA into mammalian cells.

TransIT®-2020 affords high levels of gene expression in typically hard-to-transfect cell types, including primary cells and is suitable for both transient and stable transfection. TransIT®-2020 is composed of animal-origin free components and is serum compatible, eliminating the need for any culture medium change after transfection.

Features and Benefits

  • Broad Spectrum DNA Delivery – Achieve high expression in many cell types, including hard to transfect cell lines and primary cells
  • Gentle to Your Cells – Balances high efficiency nucleic acid delivery and low cellular toxicity
  • Animal Origin Free – High performance with maximum compatibility

Other Notes

Animal Orgin Free.

Suitable for:

  • Stem cell transfection
  • RNAi gene silencing
  • Low toxicity transfection
  • Co-transfection

Legal Information

TransIT is a registered trademark of Mirus Bio




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library