Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 ea | Please contact Customer Service for Availability | $192.00 |
About This Item
$192.00
material
polystyrene
sterility
sterile
packaging
pack of 100 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio P8208
capacity
48 mL , 8 channels, max 6mL per channel
General description
Features and Benefits
- 8 or 12 channel configurations
- Compatible with Multichannel pipettors
- Individually wrapped in easy tear bags
- Sterile; RNase, DNase and pyrogen free
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description capacity 48 mL , 8 channels, max 6mL per channel, holds up to 8 reagents, sterile | description capacity 48 mL , 12 channels, max 4 mL per channel, holds up to 12 reagents, sterile | description capacity 10 mL, sterile, pack of 100 ea (individually wrapped) | description capacity 50 mL, sterile, pack of 5 ea |
| manufacturer/tradename MTC™ Bio P8208 | manufacturer/tradename MTC™ Bio P8212 | manufacturer/tradename MTC™ Bio P8010-1S | manufacturer/tradename MTC™ Bio P8050-LID |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| capacity 48 mL , 8 channels, max 6mL per channel | capacity 48 mL , 12 channels, max 4 mL per channel | capacity 10 mL | capacity 50 mL |
| material polystyrene | material polystyrene | material - | material white , polystyrene |
| packaging pack of 100 ea (individually wrapped) | packaging pack of 100 ea (individually wrapped) | packaging pack of 100 ea (individually wrapped) | packaging pack of 5 ea |
Still not finding the right product?
Explore all of our products under MTC™ Bio multichannel solution reservoir
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



