Skip to Content
MilliporeSigma

MTCP8212

MTC™ Bio multichannel solution reservoir

capacity 48 mL , 12 channels, max 4 mL per channel, holds up to 12 reagents, sterile

Synonym(s):

MTC Bio P8212, liquid reservoir, reagent holder, solution tank, solution vessel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121522
eCl@ss:
32049090
Manufacturer/tradename:
MTC™ Bio P8212
Capacity:
48 mL , 12 channels, max 4 mL per channel
Material:
polystyrene
Sterility:
sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

sterile

packaging

pack of 100 ea (individually wrapped)

manufacturer/tradename

MTC™ Bio P8212

capacity

48 mL , 12 channels, max 4 mL per channel

General description

12-Channel, 48mL, holds up to 12 reagents (12 x 4.0mL), sterile, PS, individually wrapped, 100/cs
These disposable polystyrene reservoirs are ideal for handling multiple solutions at once. Designed for use with multichannel pipettors, they are also useful on automated and semi-automated platforms and in other high-throughput applications. Individually wrapped, sterile, supplied in easy to open bags, and certified RNAse, DNase and pyrogen free.

Features and Benefits

  • 8 or 12 channel configurations
  • Compatible with Multichannel pipettors
  • Individually wrapped in easy tear bags
  • Sterile; RNase, DNase and pyrogen free

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library