Skip to Content
MilliporeSigma

MTCP827010

MTC™ Bio Serological Pipette

capacity 10 mL, purple, graduations, cotton plugged (with subdivisions), sterile, case of 200 ea (individually wrapped)

Synonym(s):

dispensing pipette, dosing pipette, MTC Bio P8270-10, dispensing tool, liquid dispensing device

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121500
Capacity:
10 mL
Sterility:
sterile
Packaging:
case of 200 ea (individually wrapped)
Feature:
graduations cotton plugged (with subdivisions)
Manufacturer/tradename:
MTC™ Bio P8270-10
Material:
polystyrene , purple
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene , purple

sterility

sterile

feature

graduations cotton plugged (with subdivisions)

packaging

case of 200 ea (individually wrapped)

manufacturer/tradename

MTC™ Bio P8270-10

technique(s)

cell culture: suitable

capacity

10 mL

General description

MTC Bio serological pipettes are made from virgin polystyrene in a class 100,000 clean room facility, under ISO 13485. Available in six volumes (1, 2, 5, 10, 25, 50ml), they are sterilized via E-beam and individually packaged in paper/plastic pouches. All sizes have vivid black, positive and negative graduations and are color coded for easy volume identification.

Features and Benefits

  • Sterile, individually wrapped
  • Ascending & descending graduations
  • Cotton plugged
  • Standardized color coding
  • Paper/plastic packaging

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?

Explore all of our products under MTC™ Bio Serological Pipette




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library