Skip to Content
MilliporeSigma

Skip To

SMB00904

Uridine 5′-diphosphoxylose sodium salt

Synonym(s):

Uridine 5′-diphosphoxylose sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
$149.00

About This Item

Empirical Formula (Hill Notation):
C14H20N2O16P2Na2
Molecular Weight:
580.24
NACRES:
NA.51
UNSPSC Code:
12352201
Assay:
≥95% (HPLC)
Form:
powder

$149.00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

powder

shipped in

wet ice

storage temp.

2-8°C

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
01897A2754I4375
description

-

description

≥90%

description

bacterial, ≥95% (HPLC)

description

≥96%

form

powder

form

powder or crystals

form

powder

form

powder

assay

≥95% (HPLC)

assay

≥90%

assay

≥95% (HPLC)

assay

≥96%

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

-

disaccharide Uridine 5′-diphosphoxylose sodium salt

-

-

-


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters