Skip to Content
MilliporeSigma

SML4011

BPTPE

≥98% (HPLC)

Synonym(s):

1,1-Bis(4-hydroxyphenyl)-2-phenyl-1-butene, 4,​4′-​(2-​Phenyl-​1-​butenylidene)​bis-phenol, 4,4′-(2-Phenylbut-1-ene-1,1-diyl)diphenol; triphenylethylene bisphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H20O2
CAS Number:
Molecular Weight:
316.39
UNSPSC Code:
51111800
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Potent partial estrogen agonist.



BPTPE, an angular estrogen, is a potent partial estrogen agonist. It induces cell cycle regulated genes that are like those activated by E2 in MCF7 cells. BPTPE exhibits delayed induction of unfolded protein response (UPR) and apoptosis in numerous breast cancer models.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library