Skip to Content
MilliporeSigma

SML4026

ZH8651 hydrochloride

≥98% (HPLC)

Synonym(s):

2-(4-Bromophenyl)ethan-1-amine hydrochloride, 4-Bromobenzene-ethanamine hydrochloride, 4-Bromophenethylamine hydrochloride, p-Bromophenethylamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H10BrN ·HCl
CAS Number:
Molecular Weight:
236.54
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

BrC1=CC=C(C=C1)CCN.Cl

InChI

1S/C8H10BrN.ClH/c9-8-3-1-7(2-4-8)5-6-10;/h1-4H,5-6,10H2;1H

InChI key

ZHAWIPAHUYMLDV-UHFFFAOYSA-N

Biochem/physiol Actions

Orally available, potent, and selective agonist of Trace amine-associated receptor 1 (TAAR1); dual activator of Gs and Gq in mTAAR1/hTAAR1.



ZH8651 is an orally available, potent, and selective agonist of Trace amine-associated receptor 1 (TAAR1). ZH8651 is dual activator of Gs and Gq in mTAAR1/hTAAR1 (EC50 = 9,85 nM (Gs), 66.5 nM Gq). ZH8651 alleviates MK-801-induced schizophrenia-like phenotypes without inducing catalepsy in mice.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library