Skip to Content
MilliporeSigma

SML4029

NVS-STG2

≥98% (HPLC)

Synonym(s):

Methyl 4-((4-(2-(tert-butyl)-5,5-dimethyl-1,3-dioxan-2-yl)benzyl)amino)-3-methoxybenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H33NO5
CAS Number:
Molecular Weight:
427.53
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C1=CC=C(NCC2=CC=C(C3(C(C)(C)C)OCC(C)(C)CO3)C=C2)C(OC)=C1)=O

InChI key

CZSPKEVHLRWAFC-UHFFFAOYSA-N

Biochem/physiol Actions

Potent STING allosteric agonist that induces a STING-mediated immune responses in cells.

NVS-STG2 is a potent STING (stimulator of interferon genes) allosteric agonist (AC=5.2 µM) that induces a STING-mediated immune responses in cells. It elicits potent antitumor activity in animal models. NVS-STG2 acts as molecular glue that binds the transmembrane domains of the neighboring STING dimers.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library