Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
0.5 mL | Please contact Customer Service for Availability | $1,110.00 |
About This Item
NACRES:
NA.32
UNSPSC Code:
12352202
Assay:
≥95% (HPLC)
Form:
aqueous solution
Mol wt:
2723.2 kDa
$1,110.00
Please contact Customer Service for Availability
assay
≥95% (HPLC)
form
aqueous solution
mol wt
2723.2 kDa
packaging
pkg of 5 mL
shipped in
dry ice
storage temp.
−20°C
General description
The Bromodomain Non-acetylated Ligand 2 is peptide histone H3 (1-21). It contains an C-terminal GG linker, followed by a biotinylated Lys. Bromodomains function as acetyl-lysine binding domains, and bromodomain/acetyl-lysine recognition can regulate histone modification, DNA replications, chromatin remodeling, and gene transcription. This peptide is suitable for use as a negative control for several bromodomains including ATAD2 and BAZ2B.
1 of 1
This Item | |||
|---|---|---|---|
| description ≥95% (HPLC) | description ≥95% (HPLC) | description 250 μg/mL, affinity isolated antibody | description 1 mg/mL, affinity isolated antibody |
| assay ≥95% (HPLC) | assay ≥95% (HPLC) | assay - | assay - |
| form aqueous solution | form aqueous solution | form buffered aqueous glycerol solution | form buffered aqueous glycerol solution |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| mol wt 2723.2 kDa | mol wt 2765.3 kDa | mol wt - | mol wt - |
| shipped in dry ice | shipped in dry ice | shipped in wet ice | shipped in wet ice |
| packaging pkg of 5 mL | packaging pkg of 5 mL | packaging - | packaging - |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



