Skip to Content
MilliporeSigma

Z-2018

ZytoLight® SPEC FOXO1/PAX3 Dual Color Single Fusion Probe

orange to green, pkg of 0.2 mL, pkg of 0.05 mL

Synonym(s):

FISH probe, dual color FISH probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

orange to green

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC FOXO1/PAX3 Dual Color Single Fusion Probe is a direct-labeled FISH probe designed for qualitative detection of the t(2;13)(q36;q14.1) translocation of the FOXO1 and PAX3 genes in formalin-fixed, paraffin-embedded specimens, using fluorescence in situ hybridization (FISH). It is intended for use with the ZytoLight FISH-Tissue Implementation Kit.

ZytoLight® probes enable the precise identification of genetic abnormalities, including translocations, deletions, amplifications, and chromosomal aneuploidies, in formalin-fixed, paraffin-embedded (FFPE) tissue sections or cytology specimens, without the need for additional antibody-based detection.

Components

The ZytoLight SPEC FOXO1/PAX3 Dual Color Single Fusion Probe comprises:

  • ZyOrange (excitation 547 nm/emission 572 nm) labeled polynucleotides (~4.5 ng/µl), targeting sequences mapping in 13q14.11* (chr13:40,816,168-41,132,595) proximal to the FOXO1 breakpoint region;
  • ZyGreen (excitation 503 nm/emission 528 nm) labeled polynucleotides (~10.0 ng/µl), targeting sequences mapping in 2q36.1* (chr2:223,196,078-223,539,352) distal to the PAX3 breakpoint region;
  • Formamide based hybridization buffer
*according to Human Genome Assembly GRCh37/hg19

Principle

The fluorescence in situ hybridization (FISH) method enables the identification and visualization of particular nucleic acid sequences within cell samples. DNA fragments labeled with fluorescent dyes, known as FISH probes, are co-denatured with their complementary target DNA strands in the sample and then allowed to hybridize. Following hybridization, non-specific and unbound probe fragments are removed through stringency washes. The DNA is then counterstained with DAPI, and the hybridized probes are detected using a fluorescence microscope equipped with filters matched to the specific fluorochromes used to label the FISH probes.

Application

This probe is intended for use in formalin-fixed, paraffin-embedded specimens to qualitatively detect t(2;13)(q36;q14.1) translocation involving the FOXO1 and PAX3 genes, via FISH.

Analysis Note

Using suitable filter sets, the hybridization signals of the probe appear orange near the FOXO1 breakpoint region, and green away from the PAX3 breakpoint region.
  • Normal scenario: in normal cells during interphase or cells without a translocation of the respective gene regions, two separate green and orange signals are observed.
  • Aberrant scenario: gene fusion is indicated by a separate orange signal, a separate green signal, and a orange/green fusion signal.
Overlapping signals may appear yellow.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library