Skip to Content
MilliporeSigma

Z-2069

ZytoLight® SPEC ESR1/CEN 6 Dual Color Probe

green to orange, pkg of 0.05 mL, pkg of 0.2 mL

Synonym(s):

FISH probe, dual color FISH probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC ESR1/CEN 6 Dual Color Probe (PL27) is a direct-labeled FISH probe for the qualitative detection of amplifications of the human ESR1 gene and the detection of chromosome 6 alpha satellites in formalin-fixed, paraffin-embedded specimens using fluorescence in situ hybridization (FISH). The probe is intended for use with the ZytoLight FISH-Tissue Implementation Kit.

ZytoLight® probes enable the precise identification of genetic abnormalities, including translocations, deletions, amplifications, and chromosomal aneuploidies, in formalin-fixed, paraffin-embedded (FFPE) tissue sections or cytology specimens, without the need for additional antibody-based detection.

Components

The ZytoLight ESR1/CEN 6 Dual Color Probe comprises:

  • ZyGreen (excitation 503 nm/emission 528 nm) labeled polynucleotides (~10 ng/µl), targeting sequences mapping in 6q25.1* (chr6:152,083,365-152,478,947) harboring the ESR1 gene region;
  • ZyOrange (excitation 547 nm/emission 572 nm) labeled polynucleotides (~1.5 ng/µl), targeting sequences mapping in 6p11.1-q11 specific for the alpha satellite centromeric region D6Z1 of chromosome 6;
  • Formamide based hybridization buffer
*according to Human Genome Assembly GRCh37/hg19

Principle

The fluorescence in situ hybridization (FISH) method enables the identification and visualization of particular nucleic acid sequences within cell samples. DNA fragments labeled with fluorescent dyes, known as FISH probes, are co-denatured with their complementary target DNA strands in the sample and then allowed to hybridize. Following hybridization, non-specific and unbound probe fragments are removed through stringency washes. The DNA is then counterstained with DAPI, and the hybridized probes are detected using a fluorescence microscope equipped with filters matched to the specific fluorochromes used to label the FISH probes.

Application

This probe is intended for use in formalin-fixed, paraffin-embedded specimens to qualitatively detect amplifications of the human ESR1 gene and the detection of chromosome 6 alpha satellites, via FISH.

Analysis Note

Using suitable filter sets, the hybridization signals of the probe appear green at the ESR1 gene region and orange at CEN 6.
  • Normal scenario: in normal cells during interphase or cells without an amplification of the ESR1 gene region, two green and two orange signals are observed.
  • Aberrant scenario: in cells with an amplification of the ESR1 gene region, more green signals or green signal clusters are observed.
Overlapping signals may appear yellow.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library