Skip to Content
MilliporeSigma

Z-2081

ZytoLight® SPEC CDKN2A/CEN 3/7/17 Quadruple Color Probe

Gold/Red/Green/Blue, pkg of 0.2 mL, pkg of 0.05 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

, Gold/Red/Green/Blue

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC CDKN2A/CEN 3/7/17 Quadruple Color Probe (PL40, Z-2081-50/200) is a FISH probe designed for the qualitative detection of the human CDKN2A gene and the alpha-satellite regions of chromosomes 3, 7, and 17 in formalin-fixed, paraffin-embedded specimens using fluorescence in situ hybridization (FISH). This probe employs ZyGold-labeled polynucleotides for the CDKN2A gene at 9p21.3 (chr9:21,742,629-22,056,853, ~5.5 ng/µl), ZyRed for chromosome 3 (D3Z1, 3p11.1-q11.1, ~0.5 ng/µl), ZyGreen for chromosome 7 (D7Z1, 7p11.1-q11.1, ~4.5 ng/µl), and ZyBlue for chromosome 17 (D17Z1, 17p11.1-q11.1, ~12 ng/µl). It is intended for use with the ZytoLight FISH-Tissue Implementation Kit (Prod. No. Z-2028-5/-20).

Principle:

The ZytoLight® system uses fluorescence in situ hybridization (FISH) to detect and visualize specific nucleic acid sequences in FFPE specimens. Direct-labeled probes co-denature with target DNA strands for hybridization. Unbound probes are washed away, and hybridized fragments are counterstained with DAPI.The ZytoLight® SPEC CDKN2A/CEN 3/7/17 Quadruple Color Probe includes:-ZyGold: Targets the CDKN2A gene at 9p21.3 (excitation 532 nm/emission 553 nm).-ZyRed: Targets chromosome 3 (excitation 580 nm/emission 599 nm).-ZyGreen: Targets chromosome 7 (excitation 503 nm/emission 528 nm).-ZyBlue: Targets chromosome 17 (excitation 418 nm/emission 467 nm).FISH visualizes gold, red, green, and blue signals corresponding to these targets. The microscopy setup eliminates the need for antibodies, using exciter, dichroic, and barrier filters for precise excitation and emission, optimized for detecting genetic aberrations like deletions and aneuploidies in tissue sections.

Components:

Probe Composition:

  • ZyGold: Labeled polynucleotides (~5.5 ng/µl) for the CDKN2A gene region.
  • ZyRed: Labeled polynucleotides (~0.5 ng/µl) for chromosome 3.-ZyGreen: Labeled polynucleotides (~4.5 ng/µl) for chromosome 7.
  • ZyBlue: Labeled polynucleotides (~12 ng/µl) for chromosome 17.
  • Hybridization Buffer: Formamide-based hybridization buffer.
Product Sizes:-Z-2081-50: 0.05 ml (5 reactions of 10 µl each).-Z-2081-200: 0.2 ml (20 reactions of 10 µl each).Related Products: Intended for use with the ZytoLight FISH-Tissue Implementation Kit (Prod. No. Z-2028-5/-20).

Application

For the qualitative detection of the human CDKN2A gene as well as alpha-satellites of chromosomes 3, 7, and 17 by fluorescence in situ hybridization (FISH)
ZytoLight® SPEC CDKN2A/CEN 3/7/17 Quadruple Color Probe aids differential diagnosis of various cancers by detecting CDKN2A deletions/aneusomies of chromosomes 3/7/17 in FFPE tissues

Analysis Note

In normal interphase nuclei, the expected results are two gold (CDKN2A), two red (CEN 3), two green (CEN 7), and two blue (CEN 17) signals. In cases of deletion of the CDKN2A gene locus, a reduced number of gold signals will be observed. Cells with aneuploidy of chromosomes 3, 7, or 17 may show varying numbers of signals corresponding to those chromosomes.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library