Skip to Content
MilliporeSigma

Z-2084

ZytoLight® SPEC VHL/CEN 3 Dual Color Probe

green to orange, pkg of 0.2 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.2 mL, pkg of 20 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC VHL/CEN 3 Dual Color Probe (PL43) is a direct-labeled FISH probe designed for the qualitative detection of deletions involving the human VHL gene and for detecting specific sequences of chromosome 3 alpha satellites in formalin-fixed, paraffin-embedded (FFPE) specimens. It is intended for use in conjunction with the ZytoLight FISH-Tissue Implementation Kit.

Principle:

The fluorescence in situ hybridization (FISH) technique detects and visualizes specific nucleic acid sequences using fluorescently-labeled DNA fragments (FISH probes) that co-denature and hybridize with complementary target DNA strands. After stringent washing to remove unbound fragments, hybridized probes are visualized with a fluorescence microscope using specific excitation and emission filters.FISH probes co-denature and hybridize with formalin-fixed, paraffin-embedded (FFPE) targets overnight at 37°C following an initial denaturation at 75°C. Unbound probes are washed away, and DAPI counterstaining is applied. Green signals for VHL and orange signals for CEN 3 are observed through matching filters.The ZytoLight® direct labeling technology eliminates the need for antibody amplification, detecting deletions via reduced VHL signals compared to the centromere.

Components:

ZytoLight® SPEC VHL/CEN 3 Dual Color Probe comprises:

  • ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm), targeting sequences in the VHL gene region on chromosome 3 (3p25.3).
  • ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm), targeting sequences in the alpha satellite centromeric region D3Z1 of chromosome 3.
  • Formamide-based hybridization buffer.

Application

For the qualitative detection of deletions affecting the human VHL gene and alpha satellites of chromosome 3 by fluorescence in situ hybridization (FISH)
The probe is designed for qualitative detection of sequences in the VHL gene region and chromosome 3 centromere in formalin-fixed, paraffin-embedded (FFPE) tissue specimens by FISH. It is used in combination with the ZytoLight® FISH-Tissue Implementation Kit for specimen pretreatment, hybridization, and post-hybridization processing.

Analysis Note

In a normal interphase nucleus, two green signals (for VHL) and two orange signals (for CEN 3) are expected. If deletions occur in the VHL gene region, a reduced number of green signals will be observed. Variations in signal patterns may indicate aberrations and should be investigated further.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library