Skip to Content
MilliporeSigma

Z-2090

ZytoLight® SPEC MYC Dual Color Break Apart Probe

green to orange, pkg of 0.2 mL, pkg of 0.05 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC MYC Dual Color Break Apart Probe (PL49, Z-2090-50/200) is a FISH probe designed for the qualitative detection of translocations involving the human MYC gene at 8q24.21 in cytologic and formalin-fixed, paraffin-embedded (FFPE) specimens, such as diffuse large B-cell lymphoma (DLBCL) and Burkitt lymphoma (BL). This probe utilizes a green probe targeting the distal region (chr8:130,373,051-130,930,673, ~10.0 ng/µl ZyGreen) and an orange probe for the proximal region (chr8:127,888,765-128,363,281, ~4.5 ng/µl ZyOrange). It is intended for use with the ZytoLight FISH Implementation Kits (Prod. No. Z-2028-5/-20 or Z-2099-20).

Principle:

The ZytoLight system uses fluorescence in situ hybridization (FISH) to detect and visualize specific nucleic acid sequences. Direct-labeled probes co-denature with their complementary target DNA strands, allowing hybridization. Unbound probes are washed away, and hybridized fragments are counterstained with DAPI, visualized using fluorescence microscopy with appropriate filters. The ZytoLight® SPEC MYC Dual Color Break Apart Probe includes: ZyGreen: Targets sequences distal to the MYC breakpoint at 8q24.21 (excitation 503 nm/emission 528 nm). ZyOrange: Targets sequences proximal to the MYC breakpoint at 8q24.21 (excitation 547 nm/emission 572 nm).FISH visualizes green and orange signals corresponding to the MYC gene regions, optimized for detecting 8q24.21 genetic aberrations in tissue sections and cytology specimens, enabling translocation detection without the need for antibodies.

Components:

Probe Composition:

  • ZyGreen: Labeled polynucleotides (~10.0 ng/µl) targeting sequences distal to the MYC breakpoint region.
  • ZyOrange: Labeled polynucleotides (~4.5 ng/µl) targeting sequences proximal to the MYC breakpoint region.
  • Hybridization Buffer: Formamide-based hybridization buffer.
Product Sizes:-Z-2090-50: 0.05 ml (5 reactions of 10 µl each).-Z-2090-200: 0.2 ml (20 reactions of 10 µl each).Related Products: Intended for use with the ZytoLight FISH Implementation Kits (Prod. No. Z-2028-5/-20 or Z-2099-20).

Application

For the qualitative detection of translocations involving the human MYC gene at 8q24.21 by fluorescence in situ hybridization (FISH)
The probe is designed for qualitative detection of translocations involving the 8q24.21 region in cytologic or formalin-fixed, paraffin-embedded (FFPE) tissue specimens by FISH. It supports both cytology and tissue workflows using compatible ZytoLight® Implementation Kits.

Analysis Note

In normal interphase nuclei, two orange/green fusion signals are expected, indicating non-rearranged loci. In cases of translocation, a signal pattern with one orange signal and one separate green signal will be observed.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library