Skip to Content
MilliporeSigma

Z-2093

ZytoLight® SPEC ERBB2/TOP2A/CEN 17 Triple Color Probe

green to orange, Blue, pkg of 0.05 mL, pkg of 0.2 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange, Blue

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC ERBB2/TOP2A/CEN 17 Triple Color Probe (PL52) is a direct-labeled FISH probe designed for the qualitative detection of amplifications involving the human ERBB2 and TOP2A genes, as well as the detection of chromosome 17 alpha satellites in formalin-fixed, paraffin-embedded (FFPE) specimens. It is intended to be used with the ZytoLight FISH-Tissue Implementation Kit.

Principle:

The fluorescence in situ hybridization (FISH) technique enables the detection and visualization of specific nucleic acid sequences. Fluorescently-labeled DNA fragments (FISH probes) co-denature and hybridize with complementary target DNA strands, followed by stringent washing to remove unbound fragments. The hybridized probes are visualized using a fluorescence microscope equipped with specific excitation and emission filters for the fluorochromes.In this assay, 10 µl of the probe is applied to pretreated formalin-fixed, paraffin-embedded (FFPE) sections. The sections are denatured for 10 minutes at 75 °C, hybridized overnight at 37 °C, and then washed and examined to detect signals: green for ERBB2, orange for TOP2A, and blue for CEN 17. The use of directly labeled DNA probes streamlines the process by eliminating the need for antibody detection.

Components:

ZytoLight® SPEC ERBB2/TOP2A/CEN 17 Triple Color Probe comprises:

  • ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting the ERBB2 gene region on chromosome 17 (17q12-q21.1).
  • ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting the TOP2A gene region on chromosome 17 (17q21.1-q21.2).
  • ZyBlue labeled polynucleotides (excitation 418 nm/emission 467 nm) targeting the alpha satellite centromeric region D17Z1 of chromosome 17.
  • Formamide-based hybridization buffer.

Application

For the qualitative detection of human ERBB2 gene amplifications, human TOP2A gene amplifications and alpha satellites of chromosome 17 by fluorescence in situ hybridization (FISH)

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library