Skip to Content
MilliporeSigma

Z-2105

ZytoLight® SPEC MYC/IGH Dual Color Dual Fusion Probe

orange to green, pkg of 0.2 mL, pkg of 0.05 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

orange to green

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC MDM2/CEN 12 Dual Color Probe (PL18) is a direct‑labeled dual‑color fluorescence in situ hybridization (FISH) probe for detecting the human MDM2 gene region together with alpha‑satellite sequences of chromosome 12 in formalin‑fixed, paraffin‑embedded specimens.It combines differently labeled probe fractions targeting the MDM2 locus and chromosome 12 centromeric sequences to support visualization of copy number changes at these regions by FISH on tissue sections.

Principle:

The ZytoLight® SPEC MDM2/CEN 12 Dual Color Probe utilizes fluorescence in situ hybridization (FISH) to detect specific nucleic acid sequences in cell preparations from formalin-fixed, paraffin-embedded (FFPE) tissue. Fluorescently labeled DNA probes—ZyGreen for the MDM2 gene region (12q15) and ZyOrange for chromosome 12 alpha satellite (CEN 12, 12p11.1-q11)—co-denature with complementary target DNA, hybridize overnight, and have unbound fragments removed via stringency washing. Counterstaining with DAPI enables visualization of hybridized green (MDM2) and orange (CEN 12) signals using a fluorescence microscope with appropriate excitation/emission filters.

Components:

Probe Composition:

  • ZyGreen: Labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting sequences in 12q15 (harboring the MDM2 gene).
  • ZyOrange: Labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting sequences in 12p11.1-q11 (specific for the alpha satellite centromeric region D12Z3 of chromosome 12).
  • Hybridization Buffer: Formamide-based hybridization buffer.
Related Products: Intended for use with the ZytoLight FISH-Tissue Implementation Kit (Prod. No. Z-2028-5/-20).

Application

For the qualitative detection of the translocation t(8;14)(q24.21;q32.3) by fluorescence in situ hybridization
The probe is designed for qualitative detection of translocations between 14q32.33 and 8q24.21 regions in formalin-fixed, paraffin-embedded (FFPE) tissue specimens by FISH. It is used in combination with the ZytoLight® FISH-Tissue Implementation Kit for specimen pretreatment, hybridization, and post-hybridization processing.

Analysis Note

The performance of the probe has been validated for detecting amplifications of the MDM2 gene locus. In normal interphase cells, two green signals (representing the MDM2 gene region) and two orange signals (representing the centromeric region of chromosome 12) are expected. In cases of amplification, multiple copies of the green signal or clusters will be observed.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library