Skip to Content
MilliporeSigma

Z-2108

ZytoLight® SPEC CCND1 Dual Color Break Apart Probe

green to orange, pkg of 0.05 mL, pkg of 0.2 mL

Synonym(s):

FISH, probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC CCND1 Dual ColorBreak Apart Probe (PL65) is a probe for qualitative detection of translocations associated the human CCND1 gene at 11q13.3.

Application

It specifically assists in differentiating atypical lipomatous tumors/well-differentiated liposarcomas (ALT/WDLPS) and dedifferentiated liposarcoma (DDLPS) by identifying CDK4 amplification, a crucial marker that distinguishes these tumors from other sarcomas.

Analysis Note

  • With the use of appropriate filter sets, the hybridization signals of the probeappear green (proximal to the CCND1 breakpoint region) and orange(distal to the CCND1 breakpoint region).
  • Normal situation: In interphases of normal cells or cells without atranslocation involving the CCND1 gene region, two green/orange fusionsignals appear
  • Aberrant situation: One CCND1 gene region affected by a translocation isindicated by one separate green signal and one separate orange signal
  • Overlapping signals may appear as yellow signals.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library