Skip to Content
MilliporeSigma

Z-2109

ZytoLight® SPEC TFE3 Dual Color Break Apart Probe

green to orange, pkg of 0.2 mL, pkg of 0.05 mL

Synonym(s):

FISH probe, dual color FISH probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC TFE3 Dual Color Break Apart Probe is a directly labeled fluorescence in situ hybridization (FISH) probe designed for the qualitative detection of translocations involving the TFE3 gene at Xp11.23. The ZytoLight® system utilizes direct labeling, eliminating the need for fluorophore-coupled antibodies and streamlining the FISH workflow. For optimal results, the probe should be used in combination with the ZytoLight FISH-Tissue Implementation Kit. ZytoLight® probes enable the precise identification of genetic abnormalities—including translocations, deletions, amplifications, and chromosomal aneuploidies—in formalin-fixed, paraffin-embedded (FFPE) tissue sections or cytology specimens, without the need for additional antibody-based detection.

Components

The ZytoLight SPEC TFE3 Dual Color Break Apart Probe comprises:

  • ZyGreen (excitation 503 nm/emission 528 nm) labeled polynucleotides (~10.0 ng/µl), targeting sequences mapping in Xp11.23* (chrX:48,906,685-49,509,699) proximal to the TFE3 breakpoint region;
  • ZyOrange (excitation 547 nm/emission 572 nm) labeled polynucleotides (~4.5 ng/µl), targeting sequences mapping in Xp11.23* (chrX:48,287,169-48,792,674) distal to the TFE3 breakpoint region;
  • Formamide based hybridization buffer
*according to Human Genome Assembly GRCh37/hg19

Principle

The fluorescence in situ hybridization (FISH) method enables the identification and visualization of particular nucleic acid sequences within cell samples. DNA fragments labeled with fluorescent dyes, known as FISH probes, are co-denatured with their complementary target DNA strands in the sample and then allowed to hybridize. Following hybridization, non-specific and unbound probe fragments are removed through stringency washes. The DNA is then counterstained with DAPI, and the hybridized probes are detected using a fluorescence microscope equipped with filters matched to the specific fluorochromes used to label the FISH probes.

Application

For the qualitative detection of translocations involving the human TFE3 gene at Xp11.23 by fluorescence in situ hybridization (FISH)

Analysis Note

Using suitable filter sets, probe hybridization produces a green signal proximal to the TFE3 breakpoint and an orange signal distal to it.
Normal scenario: female cells or any cells without a TFE3 gene region translocation display two green/orange fusion signals during interphase. Male cells or cells without such a translocation show one orange/green fusion signal in interphase.
Aberrant scenario: if a translocation affects the TFE3 gene region, this is shown by one distinct green signal and one distinct orange signal.
Overlapping orange and green signals may appear yellow.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library