Skip to Content
MilliporeSigma

Z-2111

ZytoLight® SPEC BCR/ABL1 Dual Color Dual Fusion Probe

green to orange, pkg of 0.2 mL, pkg of 0.05 mL

Synonym(s):

FISH probe, dual color FISH probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC BCR/ABL1 Dual Color Dual Fusion Probe is a direct-labeled fluorescence in situ hybridization (FISH) probe designed for the qualitative detection of the t(9;22)(q34.1;q11.2) chromosomal translocation involving the BCR and ABL1 genes, commonly associated with chronic myeloid leukemia (CML). For optimal performance, the probe is intended for use with the ZytoLight® FISH-Cytology Implementation Kit.


ZytoLight® probes enable the precise identification of genetic abnormalities—including translocations, deletions, amplifications, and chromosomal aneuploidies—in formalin-fixed, paraffin-embedded (FFPE) tissue sections or cytology specimens, without the need for additional antibody-based detection.

Components

The ZytoLight SPEC BCR/ABL1 Dual Color Dual Fusion Probe comprises:

  • ZyGreen (excitation 503 nm/emission 528 nm) labeled polynucleotides (~12 ng/µl), targeting sequences mapping in 22q11.22-q11.23* (chr22:23,000,029-24,431,064) harboring the BCR gene region;
  • ZyOrange (excitation 547 nm/emission 572 nm) labeled polynucleotides (~6 ng/µl), targeting sequences mapping in 9q34.11-q34.13* (chr9:133,223,081-134,103,849) harboring the ABL1 gene region;
  • Formamide based hybridization buffer
*according to Human Genome Assembly GRCh37/hg19

Principle

The fluorescence in situ hybridization (FISH) method enables the identification and visualization of particular nucleic acid sequences within cell samples. DNA fragments labeled with fluorescent dyes, known as FISH probes, are co-denatured with their complementary target DNA strands in the sample and then allowed to hybridize. Following hybridization, non-specific and unbound probe fragments are removed through stringency washes. The DNA is then counterstained with DAPI, and the hybridized probes are detected using a fluorescence microscope equipped with filters matched to the specific fluorochromes used to label the FISH probes.

Application

For the qualitative detection of translocation t(9;22)(q34.1;q11.2) by fluorescence in situ hybridization (FISH)

Analysis Note

Using suitable filter sets, the probe’s hybridization signals display as green for the BCR gene region and orange for the ABL1 gene region.
Normal scenario: normal cells or in cells lacking a translocation involving these gene regions, two distinct signals are observed: one green and one orange during interphase.
Aberrant scenario: where gene fusion has occurred, the signal pattern consists of one green signal, one orange signal, and two combined orange/green fusion signals.
Overlapping orange and green signals may appear yellow.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Pictograms

Health hazard

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library