Skip to Content
MilliporeSigma

Z-2114

ZytoLight® SPEC BCL2/IGH Dual Color Dual Fusion Probe

orange to green, pkg of 0.2 mL, pkg of 0.05 mL

Synonym(s):

FISH probe, dual color FISH probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

orange to green

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC BCL2/IGH Dual Color Dual Fusion Probe is a direct-labeled fluorescence in situ hybridization (FISH) probe designed for the qualitative detection of the t(14;18)(q32.3;q21.3) translocation involving the IGH and BCL2 genes. This probe enables the identification of this specific genetic aberration in formalin-fixed, paraffin-embedded (FFPE) tissue samples. For optimal performance, this probe is intended for use with the ZytoLight® FISH-Tissue Implementation Kit.

ZytoLight® probes enable the precise identification of genetic abnormalities—including translocations, deletions, amplifications, and chromosomal aneuploidies—in formalin-fixed, paraffin-embedded (FFPE) tissue sections or cytology specimens, without the need for additional antibody-based detection.

Components

The ZytoLight SPEC BCL2/IGH Dual Color Dual Fusion Probe comprises:

  • ZyOrange (excitation 547 nm/emission 572 nm) labeled polynucleotides (~6 ng/µl), targeting sequences mapping in 18q21.33* (chr18:60,180,078-61,507,691) harboring the BCL2 gene region;
  • ZyGreen (excitation 503 nm/emission 528 nm) labeled polynucleotides (~12 ng/µl), targeting sequences mapping in 14q32.33* (chr14:105,462,169-106,995,000) harboring the IGH locus;
  • Formamide based hybridization buffer
*according to Human Genome Assembly GRCh37/hg19

Principle


The fluorescence in situ hybridization (FISH) method enables the identification and visualization of particular nucleic acid sequences within cell samples. DNA fragments labeled with fluorescent dyes, known as FISH probes, are co-denatured with their complementary target DNA strands in the sample and then allowed to hybridize. Following hybridization, non-specific and unbound probe fragments are removed through stringency washes. The DNA is then counterstained with DAPI, and the hybridized probes are detected using a fluorescence microscope equipped with filters matched to the specific fluorochromes used to label the FISH probes.

Application

This probe is intended for use in formalin-fixed, paraffin-embedded specimens to qualitatively detect translocation t(14;18)(q32.3;q21.3) involving the human IGH and BCL2 genes, via FISH.

Analysis Note

Using suitable filter sets, the probe’s hybridization signals are seen as orange for the BCL2 gene region and green for the IGH locus.
  • Normal scenario: In interphase nuclei of cells without translocations affecting these gene regions, two distinct green signals and two distinct orange signals are observed.
  • Aberrant scenario: The presence of a gene fusion is shown by one separate orange signal, one separate green signal, and two orange/green fusion signals.
Overlapping orange and green signals may appear yellow.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library