Skip to Content
MilliporeSigma

Z-2175

ZytoLight® SPEC YWHAE Dual Color Break Apart Probe

green to orange, pkg of 0.05 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight SPEC YWHAE Dual Color Break Apart Probe (PL134) is a direct-labeled FISH probe designed for the qualitative detection of translocations involving the human YWHAE gene at 17p13.3 in formalin-fixed, paraffin-embedded (FFPE) specimens. It is intended to be used in combination with the ZytoLight FISH-Tissue Implementation Kit.

Components:

ZytoLight® SPEC YWHAE Dual Color Break Apart Probe comprises:
  • ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting sequences proximal to the YWHAE breakpoint region on -chromosome 17 (17p13.3).
  • ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting sequences distal to the YWHAE breakpoint region on chromosome 17.
  • Formamide-based hybridization buffer.
Principle:
FISH uses direct-labeled probes flanking YWHAE breakpoint that co-denature with target DNA in pretreated FFPE sections (10 µl/slide, sealed), hybridize overnight (75°C/10 min denaturation, 37°C hybridization), then undergo stringent washing and DAPI counterstaining for visualization via fluorescence microscopy per ZytoLight FISH-Tissue Implementation Kit (Z-2028-5/-20).

Application

ZytoLight® SPEC YWHAE Dual Color Break Apart Probe aids in the differential analysis of conditions associated with YWHAE rearrangements.. It is used for the qualitative detection of translocations involving the human YWHAE gene at 17p13.3 by fluorescence in situ hybridization (FISH).

Analysis Note

In a normal interphase nucleus, two green and two orange signals are expected. In cases of translocation, the presence of separate green and orange signals will indicate an alteration. Any unexpected signal patterns should warrant further investigation to confirm genetic rearrangements.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library