Skip to Content
MilliporeSigma

Z-2177

ZytoLight® SPEC BCL6 Dual Color Break Apart Probe

green to orange, pkg of 0.2 mL, pkg of 0.05 mL

Synonym(s):

FISH, probe

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC BCL6 Dual ColorBreak Apart Probe (PL136) is a probe for the qualitative detection oftranslocations involving the human BCL6gene at 3q27.3.
The fluorescence in situ hybridization (FISH) technique allows for the detection and visualization of specific nucleic acid sequences in cell preparations. Fluorescently-labeled DNA fragments, so called FISH probes, and their complementary target DNA strands in the preparations are codenatured and subsequently allowed to anneal during hybridization. Afterwards, unspecific and unbound probe fragments are removed by stringency washing steps. After counterstaining the DNA with DAPI, hybridized probe fragments are visualized using a fluorescence microscope equipped with excitation and emission filters specific for the fluorochromes with which the FISH probe fragments have been directly labeled.
ZytoLight SPEC BCL6 Dual Color Break Apart Probe is composed of:

  • ZyGreen (excitation 503 nm/emission 528 nm) labeled polynucleotides (~10 ng/µl), which target sequences mapping in3q27.3* (chr3:186,737,897-187,403,834) proximal to the BCL6 breakpoint region
  • ZyOrange (excitation 547 nm/emission 572 nm) labeled polynucleotides (~4.5 ng/µl), which target sequences mapping in3q27.3-q28* (chr3:187,744,962-188,097,195) distal to the BCL6 breakpoint region
  • Formamide based hybridization buffer

Application

This probe is utilized in formalin-fixed, paraffin-embedded specimens, such as in the evaluation of B-cell lymphomas and for fluorescence in situ hybridization (FISH) techniques.

Analysis Note

  • Hybridization signals of the probe appear green (proximal to the BCL6 breakpoint region) and orange (distal to the BCL6 breakpoint region)
  • Normal situation: In interphases of normal cells or cells without atranslocation involving the BCL6 gene region, two green/orange fusion signals appear
  • Aberrant situation: One BCL6 gene region affected by a translocation is indicated by one separate green signal and one separate orange signal
  • Overlapping signals may appear as yellow signals
  • Red blood cells present in the specimen might exhibit autofluorescence which hinders signal recognition

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library