About This Item
UNSPSC Code:
41122101
Material:
polyactide (PLA)
Manufacturer/tradename:
Diversified Biotech PSPD-1000
Packaging:
pack of 375 ea
material
polyactide (PLA)
feature
lid
packaging
pack of 375 ea
manufacturer/tradename
Diversified Biotech PSPD-1000
W × H
100 mm × 15 mm
General description
Planet-Safe® Cell Culture dishes are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane
Application
cell culture
Legal Information
Planet-Safe is a registered trademark of Diversified Biotech, Inc.
Choose from one of the most recent versions:
Already Own This Product?