Skip to Content
MilliporeSigma

Z744317

Planet-Safe® Cell Culture Microplates

wells, 96, round bottom, sterile, pack of 100 ea (20 bags of 5 plates)

Synonym(s):

Diversified Biotech PLATE-2000, cell cultre vessel, culture dish, laboratory dish, tissue culture plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121800
Material:
clear , polyactide (PLA) , round bottom
Sterility:
sterile
Feature:
lid: no, wells 96
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , polyactide (PLA) , round bottom

sterility

sterile

feature

lid: no, wells 96

packaging

pack of 100 ea (20 bags of 5 plates)

manufacturer/tradename

Diversified Biotech PLATE-2000

General description

100% Plant-Based Planet-Safe® 96-Well Flat bottom Microplates are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane. PLA is considerably more hydrophilic than petrochemical resins such as polystyrene. This beneficial property allows a variety of anchorage-dependent eukaryotic cell species to grow in liquid culture on the dish surface without added coatings or treatments. E-Beam Sterilized.

Application

Cell Culture

Features and Benefits

  • Derived from 100% renewable resources
  • Designed for precision and sustainability
  • Plant-based PLA composition promotes strong cell adhesion without coating, improving seeding uniformity and reproducibility.
  • Combines performance, convenience, and an 84 % reduction in life-cycle emissions, supporting environmentally responsible research practices.

Legal Information

Planet-Safe is a registered trademark of Diversified Biotech, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library