Skip to Content
MilliporeSigma

Skip To

Z745205

TWD BioX Tissue Culture Plates

48 well, flat bottom, polystyrene, sterile, individually wrapped

Synonym(s):

48-well plate, cell culture plate, individually wrapped plate, sterile plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
$336.00

About This Item

UNSPSC Code:
41122102
Material:
polystyrene
Size:
48 wells
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface

$336.00


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

sterile

packaging

case of 100

manufacturer/tradename

TWD Trade Winds Inc. BX076048

size

48 wells

binding type

Tissue Culture (TC)-treated surface

General description

BioX Sterile Tissue Culture TC-treated plates with coded wells for easy identification. Made of Polystyrene.

Application

Cell Culture

Features and Benefits

  • Smooth surface free from striation to maximize growth area
  • Raised rims on the wells with uniform rings on the lid to minimize evaporation
  • Single-position lid reduces risk of cross-contamination and handling errors
  • Wells are labelled with alphanumeric code for easy identification
  • Individually wrapped, sterile

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z745207Z745202Z745206
description

48 well, flat bottom, polystyrene, sterile, individually wrapped

description

96 well, round bottom, polystyrene, sterile, individually wrapped

description

6 well, flat bottom, polystyrene, sterile, individually wrapped

description

96 well, flat bottom, polystyrene, sterile, individually wrapped

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

manufacturer/tradename

TWD Trade Winds Inc. BX076048

manufacturer/tradename

TWD Trade Winds Inc. BX076096U

manufacturer/tradename

TWD Trade Winds Inc. BX076006

manufacturer/tradename

TWD Trade Winds Inc. BX076096

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

material

polystyrene

material

polystyrene

material

polystyrene

material

polystyrene

size

48 wells

size

96 wells

size

6 wells

size

96 wells

packaging

case of 100

packaging

case of 100

packaging

case of 18

packaging

case of 100


Still not finding the right product?

Explore all of our products under TWD BioX Tissue Culture Plates




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library