MilliporeSigma
Blood Typing Finished Reagents
Dropper bottles containing Bioscot<sup>®</sup> Blood Typing finished reagents
Finished Reagents
Millipore® portfolio of blood typing reagents are made from human or murine monoclonal antibodies and are formulated in low-protein diluents. These monoclonal reagents offer many advantages over polyclonal-based products.
  • Highly specific – no risk of contaminating antibodies
  • Lot-to-lot consistency
  • Low biohazard risk
  • IgM antibodies allow direct agglutination, react without the need for an indirect antiglobulin test, and can be used to type cells with a positive direct antiglobulin test.
We offer a large portfolio of finished reagents, designed to meet diverse commercial needs. Our premium BIOSCOT® reagents for in vitro diagnostic (IVD) use are fully compliant with EU regulation 2017/746, ensuring the highest standards of quality and reliability. Additionally, we provide finished reagents in bulk formats, reagents for research use, and unlabeled vials.
We have many years of experience in aiding customers in the registration of products in other countries, so whether your product is Millipore® labelled or in your own trade dress, we can support you with your product registration.
BIOSCOT® labelled reagents are not available to customers in all geographies.
Bulk reagent, reagent for research and unlabeled vial formats share the same reactivity and attributes as our BIOSCOT® CE marked reagents. Bulk reagent & unlabeled vial formats are for further manufacturing or non-diagnostic use. Reagents labelled for research use are not intended for use in clinical diagnostic applications.
Products
Sort by Relevance
Showing 1-11 of 11
Change View
Showing 1-11 of 11
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-11 of 11 results
Page 1 of 1
Page 1 of 1
Related Resources
Related Videos
The image depicts a professional setting where a person is wearing a white lab coat and bright blue latex gloves. They are engaged in a precise task, handling equipment or conducting an experiment in a laboratory environment filled with various blurred apparatuses in the background.
Blood Typing Monoclonal Antibodies
Explore our range of monoclonal antibodies in various forms, from standardized intermediates, FFMU, finished reagents in bulk, antibodies and reagents for research use.