Cytiva®
Cytiva® is a leading global Life Science company that provides a comprehensive portfolio of tools and technologies designed to empower researchers and biopharma companies at every stage, from discovery to delivery. Renowned for setting the gold standard in chromatography, Cytiva® resins are integral to 75% of FDA-approved biotherapeutic manufacturing processes. Choose from trusted resins like MabSelect® and streamline your chromatography workflows with AKTA® systems that offer automation and efficiency. Additionally, explore the industry's most diverse portfolio of filtration solutions with Whatman® and Pall®. With Amersham®, you can take the guesswork out of western blotting. Join us in advancing science and improving lives—together, we can drive breakthroughs that make a meaningful difference in the world.
Products
- Page 1
- Page 2
- Page 3
- Page 4
- Page 5
- Page 6
- Page 7
- Page 8
- Page 9
- Page 10
- Page 11
- Page 12
- Page 13
- Page 14
- Page 15
- Page 16
- Page 17
- Page 18
- Page 19
- Page 20
- Page 21
- Page 22
- Page 23
- Page 24
- Page 25
- Page 26
- Page 27
- Page 28
- Page 29
- Page 30
- Page 31
- Page 32
- Page 33
- Page 34
- Page 35
- Page 36
- Page 37
- Page 38
- Page 39
- Page 40
- Page 41
- Page 42
- Page 43
- Page 44
- Page 45
- Page 46
- Page 47
- Page 48
- Page 49
- Page 50
- Page 51
- Page 52
- Page 53
- Page 54
- Page 55
- Page 56
- Page 57
- Page 58
- Page 59
- Page 60
- Page 1
- Page 2
- Page 3
- Page 4
- Page 5
- Page 6
- Page 7
- Page 8
- Page 9
- Page 10
- Page 11
- Page 12
- Page 13
- Page 14
- Page 15
- Page 16
- Page 17
- Page 18
- Page 19
- Page 20
- Page 21
- Page 22
- Page 23
- Page 24
- Page 25
- Page 26
- Page 27
- Page 28
- Page 29
- Page 30
- Page 31
- Page 32
- Page 33
- Page 34
- Page 35
- Page 36
- Page 37
- Page 38
- Page 39
- Page 40
- Page 41
- Page 42
- Page 43
- Page 44
- Page 45
- Page 46
- Page 47
- Page 48
- Page 49
- Page 50
- Page 51
- Page 52
- Page 53
- Page 54
- Page 55
- Page 56
- Page 57
- Page 58
- Page 59
- Page 60
Chromatography
Researchers worldwide rely on AKTA® systems and Cytiva® prepacked columns and resins for the isolation and purification of target molecules. No matter the molecule or workflow, Cytiva® has a solution tailored for every step. Our offerings include:
- Affinity Resins: Choose from trusted options like MabSelect®, Protein Select®, His-tag, GST-tag, Strep-tag, MBP tagged resins, Protein A, Protein G, and Protein L.
- Size Exclusion Resins: Utilize our reliable Sepharcyl®, Superose®, and Superdex® resins.
- Ion Exchange Techniques: Leverage Capto® and SOURCE® for efficient separation.
- Hydrophobic Interaction Resins: Explore options such as Octyl, Phenyl, and Butyl.
Filtration
Cytiva® proudly offers the industry's most comprehensive selection of filtration devices and membranes through Whatman® and Pall®.
- Whatman® Solutions: Backed by 250 years of expertise, Whatman® products are designed to work effectively, every time. Choose from a variety of membranes, including cellulose, glass, quartz, nylon, PVDF, PTFE, and PES. Explore our filtration devices like Uniflo™ and Puradisc™ syringe filters, as well as Uniprep™ syringeless filters.
- Pall® Solutions: With Pall® now part of our portfolio, you gain access to both foundational and cutting-edge Life Science filtration solutions. The Acrodisc® syringe filter is a trusted choice, and you can select from high-quality membranes such as Supor®, Nylaflo®, Versapor®, Metricel®, Emfab®, TissuQuartz™, PVC, PTFE, and glass. Additionally, rely on the extensive Pall® filter funnel portfolio for your microbiology and testing needs, and concentrate your samples with Nanosep®, Microsep®, Macrosep®, and Jumbosep® centrifugal filters.
Western Blotting
Amersham®, the pioneer of enhanced chemiluminescence (ECL) detection, continues to innovate its portfolio to meet all your western blotting needs. Choose ECL Prime for the most sensitive ECL system available. Utilize DualVue™ and Rainbow markers to ensure accurate and precise results. Amersham® also offers high-quality nitrocellulose and PVDF blotting membranes with pore sizes ranging from 0.1 to 0.45 µm. Finally, employ Pharmalyte® carrier ampholytes for isoelectric focusing across a pH gradient, allowing for the effective separation of charged molecules (proteins or peptides) based on their isoelectric point (pI).
Gradient Density Media
Ficoll® and Percoll® are globally recognized standards for mediums designed to isolate mononuclear cells, viruses, and subcellular particles through gradient density centrifugation.
Magnetic Beads
Elevate your assays and purification methods with Sera-Mag® SpeedBeads. These amine-blocked magnetic particles are uniform, colloidally stable, mono-dispersed, and non-porous super-paramagnetic spheres, produced using a proprietary core-shell method. The core consists of a carboxylate-modified particle made through free radical emulsion polymerization of styrene and acid monomer. Magnetite (Fe3O4) is coated onto this core particle and encapsulated with proprietary polymers, while the final surface is treated with a specialized method to minimize nonspecific protein binding.
Genomics
Cytiva® offers a rapid suite of tools for working with nucleic acids. Use Illustra® kits for extraction, along with NAP®, Sephadex®, and RNAspin columns for nucleic acid sample cleanup. Discover rolling circle amplification with Phi29, which provides higher fidelity amplification compared to traditional PCR methods.
Related Resources
- Article: RNA isolation from blood, cultured cells, and animal or plant tissue on Whatman® FTA™ cards
This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
- Protocol: Reliable extraction of DNA from Whatman® FTA™ cards
This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.
- Article: Troubleshooting 2-D DIGE Results
Troubleshooting table for 2D DIGE results lists problems, causes, and prevention methods for future experiments.
- Protocol: Processing Small Tissue or Cell Samples using Sample Grinding Kit
Sample Grinding Kit from Cytiva is designed to disrupt cell or tissue samples for protein extraction.
- Protocol: Protease Inhibition using Protease Inhibitor Mix
Protease Inhibitor Mix from Cytiva inhibits serine, cysteine, metalloproteases, and calpain proteases and is suitable for animal and plant tissues, yeast, and bacteria.
- Protocol: Two-Dimensional Separation of Protein Samples
This page describes two-dimensional separation of protein samples with Cytiva products.
- Protocol: Troubleshooting first-dimension IEF: Ettan™ IPGphor™ 3 Isoelectric Focusing System
Troubleshooting first-dimension isoelectric focusing with Ettan IPGphor 3 Isoelectric Focusing System from Cytiva.
- Protocol: Desalting Samples Using Mini Dialysis Kit
Mini Dialysis Kit from Cytiva minimizes sample handling and loss, reducing streaking on 2-D gels from contaminants.
- Protocol: Cleaning Up Samples using 2-D Clean-Up Kit
2-D Clean-Up Kit from Cytiva is designed to prepare samples that would otherwise produce poor 2-D results.
- Protocol: Protein Determination using 2-D Quant Kit
A standard procedure for the quantitative determination of total protein concentration of a solution by the BCA method.