MilliporeSigma
HomeProductsMaterials ScienceBiomedical MaterialsPolyethylene Glycol (PEGs and PEOs)
Polyethylene Glycol (PEGs and PEOs)
A chalkboard illustration displaying the chemical structure of PEG (Polyethylene Glycol) with the formula C₂H₄O and a repeating unit represented as -[O-CH₂-CH₂]-n.
Polyethylene glycol (PEG), also known as polyethylene oxide (PEO) or poly(oxyethylene) (POE), is a synthetic, hydrophilic and biocompatible polyether. Typically, materials with molecular weight less than 20,000 g/mol are referred to as PEGs, whereas those with molecular weights above 20,000 g/mol are referred to as PEOs. These polymers are soluble in water as well as in many organic solvents, such as ethanol, acetonitrile, toluene, acetone, dichloromethane, hexane, and chloroform.
Products
Sort by Relevance
Showing 1-20 of 338
Change View
Showing 1-20 of 338
Compare
Product Number
Product Name
Product Description
molecular weight.default
Pricing
molecular weight.default
Mn 380-420
average Mn 400
molecular weight.default
average mol wt 8,000
molecular weight.default
Mn 5,000-7,000
average Mn 6,000
molecular weight.default
average Mn 250
molecular weight.default
average Mn 3,350
molecular weight.default
molecular weight.default
average Mv 100,000 (nominal)
molecular weight.default
average Mn 200
molecular weight.default
molecular weight.default
average Mv 600,000 (nominal)
molecular weight.default
molecular weight.default
molecular weight.default
Mn 285-315
average Mn 300
molecular weight.default
Mn 3,500-4,500
average Mn 4,000
Applications of PEGs
PEGs are non-toxic and commonly used in bioconjugation and surface functionalization applications, biomedical research, drug delivery, tissue engineering, as well as the food and cosmetics industries. The conjugation or non-covalent attachment of PEG polymer chains to molecules is referred to as PEGylation. The PEGylation method can enhance water biocompatibility and solubility, stability, and pharmacokinetic properties of therapeutics to improve their safety and efficiency in targeted diagnostics and drug delivery. PEG hydrogels are commonly used for the controlled release of therapeutics, cell culture scaffolds, regenerative medicine, wound healing, and tissue engineering.
PEG Derivatives and PEG Linkers
We offer a broad portfolio of well-defined PEGs with a wide range of molecular weights, end functionalities, reactivities, and polymer architectures.
  • Heterobifunctional PEGs
  • Homobifunctional PEGs
  • Monofunctional PEGs
  • PEG Dendrimers and Multi-Arm PEGS
  • PEG Copolymers
  • PEG Standards for GPC
  • PEG and Oligo Ethylene Glycol
  • Polyethylene oxide Powders
  • High Oligomer Purity PEGs