MilliporeSigma
Membrane Immunodetection Devices
A professional in a lab coat and purple gloves working with a Membrane Immunodetection device in a well-equipped laboratory, surrounded by various laboratory equipment and supplies, highlighting the precision and care involved in immunodetection processes.
Western blotting immunodetection involves the use of antibodies to detect target protein. Blotted membranes containing transferred proteins are blocked, followed by antibody incubation and detection using chemiluminescence, colorimetric, or fluorescent methods. Membrane immunodetection devices facilitate the processing of Western blots by simplifying workflows and accelerating detection times.
Products
Sort by Relevance
Showing 1-16 of 16
Change View
Showing 1-16 of 16
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-16 of 16 results
Page 1 of 1
Page 1 of 1
The Snap I.D. 2.0 protein detection system, showcasing a sleek white base with two blue adjustment knobs and a viewing screen, alongside designated areas for sample placement, embodying modern protein analysis technology
SNAP i.d.® 2.0 Protein Detection System
The SNAP i.d.® 2.0 Protein Detection System is a unique, vacuum-driven system that pulls reagents through the membrane, increasing membrane contact while eliminating dependence on diffusion or agitation. This improvement in three-dimensional reagent distribution decreases the length of time required for immunodetection. While traditional Western blotting requires 4 to 24 hours for the blocking, antibody incubation, and washing steps, the SNAP i.d.® 2.0 protocol takes only 30 minutes with no loss of signal intensity or reduction in blot quality. All immunodetection steps after protein transfer to a membrane (i.e., blocking, washing, and primary and secondary antibody incubations) can be performed with the SNAP i.d.® 2.0 System.