MilliporeSigma
HomeServicesProduct ServicesEmprove® ProgramEmprove® Filters and Single-Use Components
Emprove® Filters and Single-Use Components
Information is crucial for drug manufacturers selecting filtration and single-use technologies. Answering the industry-wide demand for increasing levels of transparency, our expanded Emprove® Filter and Single-use Components portfolio now covers all the major steps of the biopharmaceutical process. So you can move forward in your product development with confidence.
If you are interested in Extractables data or other premium content, Demo dossiers are available to download including data for custom assemblies, chemicals and consumables.
New Advanced Qualification Dossier
Your Fluid Management Closed & Single-Use 
Emprove® Dossier documentation addresses current and anticipated regulatory requirements to help speed up internal qualification procedures and accelerate your preparation of the drug approval process. While Emprove® Dossiers address families of components, Single-Use assemblies are composed of multiple components requiring extensive qualification. The Emprove® Advanced Qualification Dossier combines all component information into one comprehensive dossier to further Expedite Approval Preparation and Extend Compliance.
Content:
  • General information: Assembly item number, description, certification level & revision
  • Materials of Construction: Included in assembly drawing, component part numbers & descriptions
  • Specifications: Specific certification level specifications
  • Test Summaries and Results: General explanation of validation and testing, same for all assemblies
  • Regulatory Statements: Custom Regulatory Statement for entire assembly based on the assembly components
  • Extractables Data for Product Contact Components (when available): Surface areas, USP <665> Extractables Data including organic extractables and elemental impurities data
Read our Data Sheet to Learn More
Emprove® Dossiers for Filters and Single-use Components
All our high-quality portfolio products feature Emprove® Dossiers, providing the optimized information you need to fast track your drug to market. These include:
Supports product qualification and speeds up regulatory filing preparation. Includes content on the manufacturing process, product specifications and various qualification criteria (product validation data), regulatory statements, and more.
  • General information
  • Manufacturing flow chart
  • Product validation and qualification
  • Specifications (design and release criteria)
  • Materials of construction
  • Extractables summary**
  • Regulatory statements (Animal Origin, BPA, etc.)
Free of charge*
** Extractables testing of polymeric single-use components used in biopharmaceutical manufacturing, BioPhorum, published April 2020 & USP <665> draft, Plastic Components and Systems Used to Manufacture Pharmaceutical Drug Products and Biopharmaceutical Drug Substances and Products, published Sep 2020.​
Answers questions during quality risk assessment. Provides valuable information on how quality variability attributes are controlled.
  • Quality Self-assessments
  • Chain of custody
  • Supplier and CMO management
  • Shelf life testing and results
  • Packaging and Sterilization Validation
Free of charge
Supports process optimization and safety risk assessment with detailed extractables profile (BPOG extractables protocol and USP <665> chapters and information on elemental impurities (ICH Q3D).
  • Extractables Report**
  • Elemental impurities summary
  • Analytical procedures
** Extractables testing of polymeric single-use components used in biopharmaceutical manufacturing, BioPhorum, published April 2020 & USP <665>, Plastic Components and Systems Used to Manufacture Pharmaceutical Drug Products and Biopharmaceutical Drug Substances and Products, finalized November 2021 and effective May 2026.
Emprove® Single-Use Specific Reports 
The Advanced Qualification Dossier is customized with specific information for your assembly. The dossier can be purchase individually or subscribe to the Emprove® Suite
Supports product qualification and speeds up regulatory filing preparation. Includes content on the manufacturing process, product specifications and various qualification criteria.
  • General information
  • Manufacturing flow chart
  • Product validation and qualification
  • Specifications (design and release criteria)
  • Materials of construction
  • Regulatory statements (Animal Origin, BPA, etc.)
Supports process optimization and safety risk assessment with detailed extractables profile (BPOG extractables protocol and USP <665> chapters and information on elemental impurities (ICH Q3D).**
  • Surface Area for product contact components
  • Extractables Data for product contact components (when available)
  • Elemental impurities summary
**Extractables testing of polymeric single-use components used in biopharmaceutical manufacturing, BioPhorum, published April 2020 & USP <665>, Plastic Components and Systems Used to Manufacture Pharmaceutical Drug Products and Biopharmaceutical Drug Substances and Products, finalized Nov 2021, effective May 2026.
Support end-user risk assessment activities when particulates (particles) are found in/on Mobius® Single-Use Assemblies
  • Identified Particulates
  • Characteristics
  • FTIR Spectrum
Support end-user risk assessment for demonstration of sterility.
Supports process optimization and safety risk assessment with detailed extractables profile (USP <665> chapter and information on elemental impurities (ICH Q3D).
  • Extractables Report**
  • Elemental impurities summary
  • Analytical procedures
** USP <665>, Plastic Components and Systems Used to Manufacture Pharmaceutical Drug Products and Biopharmaceutical Drug Substances and Products, finalized November 2021 and effective May 2026.
Emprove® Filtration and Single-use Component Product Groups*
For an overview of our portfolio products and dossier availability, select any product family below. 
*Status as per Q1 2023, constantly updated.
Filter
Material Qualification Dossier
Quality Management Dossier
Operational Excellence Dossier
Pellicon® 3 Cassettes with Biomax® Membranes 
Pellicon® 3 Cassettes with Ultracel® Membranes
Pellicon® Capsules with Ultracel® Membrane 
Cellicon® Perfusion Filter 
Filter
Material Qualification Dossier
Quality Management Dossier
Operational Excellence Dossier
Viresolve® Pro Modus and Magnus Devices
Viresolve® Pro Magnus Devices
Viresolve® Pro Modus and Magnus Shield
Viresolve® Pro Modus and Magnus Shield H 
PureFlex™ Film
Material Qualification Dossier
Quality Management Dossier
*Operational Excellence Dossier
PureFlex™ Single-use Process Container Films
PureFlex™ Plus Single-use Process Container Films
Ultimus® Single-Use Process Container Film 
* Available through subscription
Lynx® ConnectorMaterial Qualification DossierQuality Management DossierOperational Excellence Dossier
Lynx® S2S Connectors
Lynx® CDR Connectors
Lynx® ST Connectors
Filter
Material Qualification Dossier
Quality Management Dossier
Operational Excellence Dossier
Milligard® PES Opticap® XLT Filters
Milligard® PES Cartridge Filters 
Milligard® PES Small Scale and T-Line Capsules
Millistak+® HC Pro Process Scale Pod Depth Filters
Clarisolv® Depth Filters 
Fast track your extractables risk assessment
All polymeric components, such as the filters and single-use systems used in biopharmaceutical manufacturing, must be assessed for safety through an evaluation of extractables and potential leachables. Extractables are compounds that can be extracted from these materials, while leachables are compounds that leach from the materials into the process stream. An understanding of extractables is essential as it can help identify the leachables that may enter a process stream.
With our Emprove® Program, developing an extractables profile for filters and single-use systems no longer requires a time- and resource-intensive compilation of a vast amount of data. Instead, our Emprove® Operational Excellence Dossiers provide you with reliable extractables data based on industry standards and guidelines established by the BioPhorum Operations Group's (BPOG) standardized testing protocol and USP <665> chapters. The datasets generated facilitate your safety risk assessment, accelerating your ability to identify potential leachables and calculate patient exposure based on process conditions, total surface area in contact with the product, batch size and final drug product maximum daily dose.
BPOG Requirements
USP <665>
(Finalized November, 2021)
Emprove® Program Approach
Scope
Single-use components in contact with fluid path (for biopharmaceutical manufacturing)
Single-use and multi-use components and devices with fluid path contact (for pharmaceutical drug product and biopharmaceutical drug substance and drug product manufacturing)
Single-use and multi-use components and devices with fluid path contact, all relevant existing and new products
Solvents
1. 50% Ethanol
2. 0.5N NaOH
3. 0.1M Phosphoric Acid
4. WFI
1. 0.2M KCl, pH 3 (C1)
2. 0.1M Phosphate buffer, pH 10 (C2)
3. 50% Ethanol (C3)
= BPOG + USP <665>
Analytical Methods
HPLC-PDA/MS (APCI, ESI, +/-)
ICP/MS, GCMS-DI, GCMS-HS (TOC, pH, NVR)
Described in USP <1663>
Broader scope in method selection
BPOG, additionally Ion Chromatography
Time Points
1-3, dependent on component
Described in USP <1663>
Broader scope in method selection
= BPOG + USP <665>
Pre-treatment
“… should be pre-treated the same way before … extractables testing…”
“… tested when they have been conditioned or processed in a manner consistent with their intended use and as specified in the manufacturer's instructions for use.”
Separate tests and reports for gamma irradiation or autoclave pre-treatment
No pre-flush unless required (worst case)
A full spectrum of services for extractables and leachables risk assessment
Together with our BioReliance® Validation Services experts, we provide a higher level of confidence in safety risk assessment through careful selection of well-characterized materials, experience in identifying and analyzing potential leachables (or process-specific extractables) data, and expertise in the design and execution of need-based leachables studies. Because every product and process is unique, we tailor our approach to deliver best-in-class services and robust results that meet global industry standards, even as the standards continue to evolve.
EMPROVE® Advanced Qualification Dossier and Component Extractables Reports
For Mobius Assemblies that contain multiple components, the Advanced Qualification Dossier includes assembly and component information to support the qualification and patient safety.  Included are the extractable datasets that are individually available through the Operational Excellence Dossiers (film, connectors, and filters) and Third-Party component Extractables Reports.  To evaluate your specific product and process, further support through consultation and services from  Validation Services are available. The Advanced Qualification Dossier or individual component Extractable reports can be obtained from the links below. 
AssemblyTest ApproachMaterials of Construction / ResinPre-treatmentAccess to Extractables Data
Mobius® Single-Use Assemblies (Full Component Evaluation)BPOG, USP <665> Multiple Components based on AssemblyGammaAdvanced Qualification Dossier
Individual Third-Party Components
BPOG, USP <665>
Component Specific
Gamma
Emprove® Component Extractables Reports
Emprove® Operational Excellence Dossiers by Product Family
A thorough interpretation of the extractables datasets from our Operational Excellence Dossiers to your specific product and process can further be supported through consultation and services from Bioreliance® Validation Services. For an overview of available Operational Excellence Dossiers, select any product family below.
* P/N stands for part number. The indicated part numbers are generic for the family. Please replace your specific part numbers as applicable.
Connector
Test Approach
Materials of Construction / Resin
Pre-treatment
Representative P/N**
Lynx® CDR Connectors
BPOG / USP Medium Risk
PS, Silicone, Stainless Steel, Glass-filled PS
Gamma
CDRXXXN05
Lynx® S2S Connectors
BPOG / USP Medium Risk
PS, Silicone
Gamma
SSCXHBXAXX
Lynx® ST Connectors
BPOG / USP Medium Risk
PS, Silicone
Gamma
STCXXXXXXX
Filter
Test Approach
Materials of Construction / Resin
Pre-treatment
Representative P/N**
Milligard® PES Capsule FiltersBPOG / USP High RiskPES, PPAutoclaveKMXXXXXXXX
Milligard® PES Cartridge Filters BPOG / USP High RiskPES, PP
GammaCMXXXXXXXX
Millistak+®  HC Pro Process Scale Pod Depth Filters (C0SP and D0SP)
BPOG / USP High Risk
PP, Silica
NA*
MC0SPXXXXX, MD0SPXXXXX
Millistak+® HC Pro Process Scale Pod Depth Filters (X0SP)
BPOG / USP High Risk
PP, Silica
NA*
MX0SPXXXXX
Process Container Film
Test Approach
Materials of Construction / Resin
Pre-treatment
Representative P/N**
PureFlex™ Plus Single-use Process Container Film
BPOG / USP Medium Risk
ULDPE, EVOH, EVA
Gamma
NA*
PureFlex™ Single-use Process Container Film
BPOG / USP Medium Risk
ULDPE, EVOH, EVA
Gamma
NA*
Ultimus® Single-Use Process Container Film BPOG / USP High RiskULDPE, EVOH, EVA GammaNA*
Sterile FilterTest ApproachMaterials of Construction / ResinPre-treatmentRepresentative P/N**
Autoclavable Opticap® XL and XLT Capsule Filters with Hydrophilic 0.1 µm Durapore® MembraneBPOG / USP Medium RiskPVDF, PP, SiliconeAutoclaveKVVLAXXXXX
Autoclavable Opticap® XL and XLT Capsule Filters with Hydrophilic 0.22 µm Durapore® MembraneBPOG / USP Medium RiskPVDF, PP, SiliconeAutoclaveKVGLAXXXXX
Hydrophilic 0.1 µm Durapore® CartridgeBPOG / USP Medium RiskPVDF, PP, SiliconeAutoclaveCVVLXXXXX
Hydrophilic 0.22 µm Durapore® CartridgeBPOG / USP Medium RiskPVDF, PP, SiliconeAutoclaveCVGLXXXXX
Autoclavable Opticap® XL and XLT Capsule Filters with Hydrophilic 0.45/0.22 µm Durapore® MembraneBPOG / USP Medium RiskPVDF, PP, SiliconeAutoclaveKHGLAXXXXX
Millipak® Final Fill 0.22 µm and 0.1 µm FiltersBPOG / USP High RiskPVDF, PSGammaMFVLXXXXX, MFGLXXXXX
Millipak® Final Fill 0.45 µm FilterBPOG / USP High RiskPVDF, PSGammaMFHLXXXXX
Millipak® Final Fill 5.0 µm FilterBPOG / USP High RiskPVDF, PSGammaMFSLXXXXX
Millidisk 0.1 µm FilterBPOG / USP Medium RiskPVDF, PSAutoclaveMCHLXXXXX
Millidisk 0.22 µm FilterBPOG / USP Medium RiskPVDF, PSAutoclaveMCGLXXXXX
Milligard® PES Capsule Filters / USP High RiskPES, PPAutoclaveKMXXXXXXXX
Milligard® PES Cartridge FiltersBPOG / USP High RiskPES, PPGammaCMXXXXXXXX
Gamma Compatible and Sterile Opticap® XL Capsule Filters with  Millipore Express® SHC, SHF, SHR, and SHR with Prefilter MembranesBPOG / USP Medium RiskPES,PET, PP, SiliconeGammaKHVEGXXXXX, KHVESXXXXX, KVEPGXXXXX, KVEPSXXXXX, KHGEGXXXXX, KHGESXXXXX, KGEPGXXXXX, KGEPSXXXXX
Gamma Compatible and Sterile Opticap® XLT Capsule Filters with Millipore Express® SHC and SHR Membrane – High AreaBPOG / USP High RiskPES,PET, PP, SiliconeGammaKHVEGXXXXX, HKHGEGXXXXXX, KHGESXXXXXX,
Gamma Compatible and Sterile Opticap® Small Scale Capsule Filters with  Millipore Express® SHC, SHF, SHR, and SHR with Prefilter MembranesBPOG / USP Medium RiskPES,PE, PP, SiliconeGammaKHVEG0XXXXX, KHVES0XXXXX, KVEPG0XXXXX, KVEPS0XXXXX, KHGEG0XXXXX, KHGES0XXXXX, KGEPG0XXXXX, KGEPS0XXXXX
Autoclavable Opticap® XL and XLT Capsule Filters with Millipore Express® SHC, SHF, SHR, and SHR with Prefilter Membranes PES,, PP, Silicone KHVEAXXXXX, KVEPAXXXXX, KHGEAXXXXX, KGEPAXXXXX
Millipore Express® SHC, SHF, SHR, and SHR with Prefilter Cartridges PES,, PP, Silicone CHVEXXXXX, CVEPXXXXX, CHGEXXXXX, CGEPXXXXX
TFF Filter
Test Approach
Materials of Construction / Resin
Pre-treatment
Representative P/N**
Pellicon® 3 Cassettes with Biomax® Membranes
BPOG / USP High Risk
PES, PP, PE
NA
P3B0XXXXX
Pellicon® 3 Cassettes with Ultracel® Membranes
BPOG / USP High Risk
Composite Regenerated Cellulose, PP, PE
NA
P3C0XXXXX
Pellicon® Capsules with Ultracel® Membrane
BPOG / USP High Risk
Composite Regenerated Cellulose, PP, PE
Gamma
PCC030CXX, PCC030CXXX
Virus Filter
Test Approach
Materials of Construction / Resin
Pre-treatment
Representative P/N**
Viresolve® Pro Modus and Magnus Devices 
BPOG / USP High Risk
PES, PVDF, Silicone
N/A
VPMDXXXXXX,
Viresolve® Pro Magnus Devices BPOG / USP High RiskPES, PVDF, SiliconeN/AVPMGXXXXXX
Viresolve® Pro Modus and Magnus Shield BPOG / USP High RiskPES, PVDF, SiliconeN/AVPPSXXXXXX
Viresolve® Pro Modus and Magnus Shield H BPOG / USP High RiskPES, PVDF, SiliconeN/AVPPHXXXXXX
The new Emprove<sup>®</sup> Suite
THE NEW EMPROVE® SUITE: SPEED THROUGH COMPLEXITY WITH CONFIDENCE
Compliance with current Good Manufacturing Practices is a continuum. Changes are inevitable. Audits and periodic reviews are necessary.
Take a seat: The Emprove® Suite is here to improve your risk assessment journey. This  Information-as-a-Service digital platform is your co-pilot to support your need for convenient access to reliable information – anywhere, anytime. A subscription helps you stay current:  You can  not only easily find, view and download Emprove® Dossiers, but also to “opt-in” to notification updates to be informed of changes to documents. You can also generate Emprove® Dossier download metrics and reports, and so much more.
Ultimately, a subscription to the Emprove® Suite empowers you to:
  • take advantage of everything the Emprove® Program has to offer
  • collaborate and share information globally
  • speed your way through the complexity of compliance with confidence
Related Product Resources
Page 1 of 2
To subscribe today, simply complete and submit the reply form below after reading our Emprove® terms and conditions.