HomeGas Chromatography (GC) OSHA Method 52: GC Analysis of Formaldehyde on SUPELCOWAX® 10 After Collection/Desorption Using ORBO™-24
OSHA Method 52: GC Analysis of Formaldehyde on SUPELCOWAX® 10 After Collection/Desorption Using ORBO™-24
Description | |
|---|---|
| Sample/matrix | HMP-coated Supelpak 20 spiked with formaldehyde (10 mug) |
| Adsorbent tube | ORBO-24 (20231) |
| Column | SUPELCOWAX 10, 30m x 0.53 mm I.D., 0.5 µm (25325) |
| Oven | 50 °C (2 min) to 200 °C at 10 °C/min |
| Carrier gas | helium, 7 mL/min |
| Injection | 1 µLdirect, 220 °C |
| Detector | NPD, 220 °C |
Description | |
|---|---|
| Categories | Analytical Chromatography, GC Applications, Air Monitoring Applications, Aldehydes & Ketones, Volatiles, Aldehydes |
| Featured Industry | Industrial Hygiene |
| General description | This is an application, not a product for sale. For more information, email: [email protected] |
| Legal Information | SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC, ORBO is a trademark of Sigma-Aldrich Co. LLC |
| Other Notes | Complementary products: Syringes Vials Gas Purification and Gas Management Inlet Liners, Glass Wool, and Specialized Hand Tools Septa and Specialized Hand Tools Column Ferrules, Nuts, and Specialized Hand Tools |
| Suitability | Application for air monitoring, application for GC |
Analyte
- 2-(Hydroxymethyl)piperidine (HMP)
- Formaldehyde derivative
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500