HomeGas Chromatography (GC) US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 µm), Fast GC Analysis
US EPA Method 8015 (modified): GC Analysis of Diesel Range Organics (DRO) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 µm), Fast GC Analysis
Conditions
| Column | SLB-5ms, 20 m x 0.18 mm I.D., 0.18 µm (28564-U) |
| Oven | 50 °C (2 min), 20 °C/min to 325 °C (1 min) |
| Carrier Gas | helium, 0.9 mL/min, constant |
| Sample | each analyte at 50 µg/mL in methylene chloride |
| Injection | 1 µL, splitless (splitter open at 0.5 min) |
| Liner | 2 mm I.D., FocusLiner™ inlet liner |
| Inj. Temp. | 250 °C |
| Detector | FID, 350 °C |
Description
| Categories | Analytical Chromatography, GC Applications, Hydrocarbons |
| Featured Industry | Environmental |
| Other Notes | Complementary products: Syringes Vials Gas Purification and Gas Management Inlet Liners, Glass Wool, and Specialized Hand Tools Septa and Specialized Hand Tools Column Ferrules, Nuts, and Specialized Hand Tools |
| Suitability | application for GC |
Analyte
- 5-α-Androstane
- Decane
- Docosane
- Dodecane
- Eicosane
- Hexacosane
- Hexadecane
- Methylene chloride
- Octacosane
- Octadecane
- o-Terphenyl
- Tetracosane
- Tetradecane
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500