MilliporeSigma
HomeSmall Molecule HPLCHPLC Analysis of Vitamins B1, B6 and B12 Using Ascentis® Express RP-Amide
HPLC Analysis of Vitamins B1, B6 and B12 Using Ascentis® Express RP-Amide
HPLC Analysis of Vitamins B1, B6 and B12 using Ascentis® Express RP-Amide
Figure 1.HPLC Analysis of Vitamins B1, B6 and B12 using Ascentis® Express RP-Amide
Conditions
ColumnAscentis Express RP-Amide, 10 cm x 4.6 mm I.D., 5 µm particles (50773-U)
Column Temp.30 °C
Mobile Phase[A] 0.02 M Ammonium Formate, pH = 3.0; [B] Acetonitrile
Gradient0-50% B in 1 min. Hold at 50% B until 1.5 min
Flow Rate1.5 mL/min
Pressure1639 psi (113 bar)
SampleDissolved in 50/50 water/ACN
Injection1 µL
DetectorUV, 276 nm
Description
CategoriesAnalytical Chromatography, Vitamins and Natural products
Featured IndustryVitamins, Nutraceuticals, and Natural Products
Legal InformationAscentis is a registered trademark of Sigma-Aldrich Co. LLC
SuitabilityApplication for HPLC
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500