MilliporeSigma
HomeSmall Molecule HPLCAnalysis: Activated Nucleotide Sugars by LC-MS/MS
Analysis: Activated Nucleotide Sugars by LC-MS/MS
Tina Wigger, Scientific Expert
MilliporeSigma
Article from Analytix Reporter - Issue 14
Introduction
Nucleotide sugars, or nucleotide activated sugars, are highly energetic forms of monosaccharides that act as key metabolites in glycosylation reactions, during which the glycosyl group from the activated sugars is transferred to an acceptor molecule, e.g. a protein.
In mammals, the most common nucleotide for sugar activation is uridine diphosphate (UDP), which is found in combination with Glucose (Glc), Galactose (Gal), N-Acetylglucosamine (GlcNAc), N-Acetylgalactosamine (GalNAc), Glucuronic acid (GlcA) and Xylose (Xyl). Furthermore, Guanosine diphosphate (GDP) linked to Mannose (Man) and Fucose (Fuc) as well as Cytidine monophosphate (CMP) linked to sialic acid (Neu5Ac) are used for glycan assembly. Plants and bacteria utilize an even larger variety of nucleotides and sugars.
Understanding the nucleotide sugar metabolism is of interest in different scientific areas, e.g. for the production of glycosylated therapeutic proteins in cell culture, where a sufficient glycosylation needs to be ensured in order to obtain a product of high quality and efficacy.1 For this purpose, a sensitive, quantification method is needed that is capable of simultaneously analyzing a set of polar analytes with similar structures and physicochemical properties. In this application note, an LC method using a Supel™ Carbon LC column in combination with selective MS/MS detection is described.
Experimental Conditions
The used conditions for the analysis of 11 nucleotide activated sugars in a standard solution with a concentration of 80 µM each are described in Table 1 & 2.
ColumnSupel™ Carbon LC, 50 x 3 mm I.D., 2.7 µm (59991-U)
Mobile phase[A] Water + 0.5 % diethylamine, pH 10 (adjusted with acetic acid); [B] acetonitrile
GradientTime (minutes)A%B%
01000
2.01000
12.59010
21.02575
26.02575
26.51000
33.01000
Flow rate0.4 mL/min
Column temp.60 °C
DetectorMS/MS, negative mode (Instrument: Agilent 6495 Triple Quadrupole); for used transitions see Table 2
Injection10 µL
Needle Wash SolutionWater + 5% isopropanol
Sample(s)10 mM individual stock solutions of the activated sugar standards were prepared in water and mixed to yield a concentration of 80 µM each.
Table 1.HPLC Conditions for analysis of 11 nucleotide activated sugars
AnalyteQ1 mass (m/z)Q3 mass (m/z)
ADP-Glucose588.1346.0
ADP-Ribose558.1346.1
CMP-Neu5Ac613.2322.0
GDP-Fucose588.1442.1
GDP-Mannose604.0424.0
UDP-Galactose565.0323.0
UDP-GalNAc/UDP-GcNAc606.1385.1
UDP-Glucose565.0323.1
UDP-Glucuronic acid579.0403.1
UDP-Xylose535.0323.1
TDP-Glucose563.1321.1
Table 2.MS/MS Transitions nucleotide activated sugars
Results
The 5 cm x 3 mm I.D. Supel™ Carbon LC column provided, under reversed phase conditions, good retention of the 11 activated nucleotide sugar compounds in the applied standard solution (Figure 1 & Table 3).
LC-MS/MS Analysis of nucleotide activated sugars on a Supel™ Carbon LC column.
Figure 1.LC-MS/MS Analysis of nucleotide activated sugars on a Supel™ Carbon LC column.
Peak no.CompoundRetention time (min)
1CMP-Neu5Ac12.75
2UDP-Glucose12.89
3UDP-Glucuronic acid12.91
4UDP-Galactose13.41
5UDP-Xylose13.49
6UDP-GalNAc13.62
7GDP-Mannose14.55
8TDP-Glucose14.65
9GDP-Fucose14.81
10ADP-Ribose15.05
11ADP-Glucose15.11
Table 3.Peak identification and retention times of nucleotide activated sugars on the Supel™ Carbon LC column
Conclusion
This application demonstrated the use and suitability of the Supel Carbon LC column for the analysis of 11 nucleotide activated sugars. The group of highly polar compounds with similar structures and physicochemical properties was sufficiently retained and separated on the carbon LC column to be selectively detected with the described MS/MS method. Its ability to retain polar compounds makes the Supel Carbon LC column a viable choice for the analysis and quantitation of nucleotide activated sugars under reversed phase (RP) conditions.
See more information about the Supel™ Carbon LC column at SigmaAldrich.com/carbonlc
Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Glycans in Biotechnology and the Pharmaceutical Industry. In: Varki A, Cummings RD, Esko JD, et al., editors. Essentials of Glycobiology. 2nd edition. Cold Spring Harbor (NY): Cold Spring Harbor Laboratory Press; 2009. Chapter 51. [Internet]. Available from: https://www.ncbi.nlm.nih.gov/books/NBK1908/