Predesigned siRNA
MISSION® Predesigned siRNA were created using the proprietary Rosetta Inpharmatics siRNA Design algorithm in an exclusive partnership with Merck & Co. The Rosetta siRNA Design Algorithm utilizes Position-Specific Scoring Matrices and knowledge of the seed region to predict the most specific and effective sequences for your target genes. The algorithm’s rules were developed utilizing empirical data collected from gene silencing experiments carried out over three years.
Product Benefits
- Best-in-class, guaranteed gene silencing
- Efficient knockdown of low abundance messages
- Simplified transfection optimization with 11 Positive Control siRNA
- Distinguish sequence-specific silencing from non–specific effects with 8 negative control siRNA
- Hundreds of functionally-validated predesigned siRNA
Product Features
- Species: Human, Mouse & Rat
- Quantities: 2 (10 nmol), 5 (25 nmol) & 10 (50 nmol) OD
- Purification: Desalt or HPLC
- Sequence Form: 21mer duplexes with dTdT overhangs
- Quality Control: 100% mass spectrometry*
- Format: Supplied dry in tubes
*Depending on manufacturing site, PAGE may be used to assess siRNA duplexes.
Predesigned Guarantee
The guarantee applies to three purchased MISSION® Predesigned siRNA targeting the same gene will reduce target mRNA levels by 75%, in cultured cells, when transfected at a concentration of ≥30 nM as a pool. If the knock down of the target gene measured between 24 to 72hours post transfection is below 75%, we will provide three additional siRNA for that gene, free of charge. If there are no more siRNA for that gene or all siRNA fail to knock down the target gene by 75%, we will refund the purchase price.
Receipt of appropriate supporting data for transfection efficiency is required for the guarantee to apply. Appropriate supporting data for transfection efficiency would include qPCR data comparing target mRNA levels of a MISSION Positive Control siRNA (GAPDH, MAPK1, TP53, etc.), transfected at ≥30 nM, to an appropriate negative control (such as mock transfection, a scrambled siRNA sequence, or MISSION Universal Negative Control siRNA), demonstrating knockdown of the target mRNA of 75%.
Due to the variability of antibodies and protein half-lives, we are unable to accept data from protein-based detection methods.
Product Pools
A popular pooled format is 4 duplexes at 5 nmol each combined into one tube (20 nmol pooled) plus the exact same 4 duplexes also at 5 nmol each in separate tubes (another 20 nmol individual). However, our sophisticated liquid handlers allow for a wide range of other possibilities. For feasibility review of your specific needs, contact [email protected].
Validated siRNA
Many common gene targets have been validated for ≥75% mRNA knockdown (Figure 1 for example data and the table for a list of commonly-ordered, validated siRNA by gene symbol). Validated siRNA are suitable for transfection optimization and as positive controls.
Figure 1.HeLa cells transfected with predesigned siRNA at a concentration of 30 nM. The percentage remaining gene expression levels were measured via qPCR 48 hours after transfection (relative to mock). Data represents the mean of four biological replicates.
| Validated siRNA by Gene Symbol | |||||||
|---|---|---|---|---|---|---|---|
| ABCC1 | CDC7 | DYRK1A | IGFBP4 | MLH1 | PIK3R4 | PRPS1 | SIK2 |
| ACP1 | CDC73 | DYRK1B | IL6ST | MSH2 | PIM2 | PRSS23 | SLC16A1 |
| ACP2 | CDK1 | EEF2K | ILK | MST4 | PIN1 | PSEN1 | SLC25A17 |
| ACVR1 | CDK11B | EIF2AK4 | IMPA2 | MTA2 | PINK1 | PSEN2 | SLC2A1 |
| ACVR1B | CDK17 | EIF4A3 | INPPL1 | MTM1 | PIP4K2B | PSMA7 | SLC30A1 |
| ADAM10 | CDK2 | EMR2 | IP6K1 | MTMR1 | PIP5K1A | PSMB4 | SLK |
| ADAM12 | CDK4 | ENPP1 | IP6K2 | MTMR2 | PKN2 | PSMB5 | SMU1 |
| ADAM15 | CDK5 | EPHA5 | IPMK | MTMR3 | PLAUR | PSMB6 | SNRK |
| ADIPOR1 | CDK6 | EPHB4 | IRAK1 | MTMR6 | PLK1 | PSPH | SORT1 |
| ADIPOR2 | CDK7 | F3 | IRAK4 | MTOR | PLK4 | PTK2 | SRC |
| ADRBK1 | CDK8 | FADD | ITPK1 | NADK | POLK | PTP4A1 | SRPK1 |
| AKT1 | CDK9 | FDFT1 | JAK1 | NCSTN | PPAP2C | PTPLA | ST7 |
| AKT2 | CDKL5 | FER | JUND | NDRG1 | PPAT | PTPN1 | STAT3 |
| ALPL | CDKN1B | FMNL1 | JUP | NEDD8 | PPM1D | PTPN11 | STK16 |
| APP | CDKN3 | FURIN | LANCL1 | NEK2 | PPME1 | PTPN12 | STK24 |
| ARAF | CELSR1 | FYN | LATS1 | NEK6 | PPP1CA | PTPN14 | STK3 |
| ARHGDIA | CERK | FZD4 | LDHA | NEK7 | PPP1CB | PTPN23 | STYX |
| ATF1 | CHEK1 | FZD5 | LEPR | NEK9 | PPP1CC | PTPN9 | TAOK1 |
| ATM | CHUK | FZD6 | LGALS3 | NET1 | PPP1R11 | PTPRE | TAOK3 |
| ATP6V0C | CKS2 | GLTSCR2 | LIMK2 | NF1 | PPP1R12A | PTPRF | TBK1 |
| ATR | CLPP | GPRC5A | LRP5 | NLN | PPP1R2 | PTPRJ | TEK |
| AURKB | CPE | GRB2 | LRP6 | NME1 | PPP1R3C | PTPRK | TESK1 |
| AXIN1 | CPT1A | GRK6 | LTBR | NME1-NME2 | PPP1R7 | PTPRS | TFRC |
| AXL | CRKL | GRN | LYN | NME2 | PPP2CA | RAB22A | TGFBR1 |
| BACE1 | CSK | GSK3B | MAD2L1 | NOTCH2 | PPP2CB | RAF1 | TGM1 |
| BACE2 | CSNK2A1 | HADHB | MANF | NPR1 | PPP2R1A | RAP1B | TGM2 |
| BAD | CTNNB1 | HBEGF | MAP2K2 | NR1H2 | PPP2R2A | RARG | THRA |
| BAG1 | CTSA | HCG 1757335 | MAP2K5 | NR1H3 | PPP2R5A | RB1 | TK1 |
| BCAT1 | CXCR4 | HDAC1 | MAP3K3 | NR2C2 | PPP2R5D | RELA | TKT |
| BIRC5 | CXCR7 | HDAC2 | MAP3K4 | NR2F2 | PPP2R5E | RHOA | TLR4 |
| BMP6 | CYLD | HECTD1 | MAPK14 | NUP85 | PPP3CC | RIOK3 | TM4SF1 |
| BMPR1A | DAD1 | HIPK2 | MAPK3 | OCRL | PPP5C | RIPK1 | TNFRSF10B |
| BRAF | DAPK3 | HIPK3 | MAPK6 | OXSR1 | PPP6C | RIPK2 | TRIM28 |
| BUB1 | DCN | HPRT1 | MAPK8 | PAK2 | PREPL | RNF10 | TWF1 |
| BUB1B | DDR1 | HSPA1A | MAPK9 | PASK | PRKACA | RNF5 | TYRO3 |
| CASK | DGKA | HSPA1B | MAPKAPK5 | PBK | PRKACB | ROCK1 | VEGFC |
| CCL2 | DMBT1 | HSPB8 | MARK3 | PCNA | PRKAG1 | ROCK2 | VIPR1 |
| CCNA2 | DNMT1 | HTRA1 | MASTL | PCSK9 | PRKAR1A | ROR2 | VRK2 |
| CCNC | DUSP10 | HUNK | MBTPS1 | PDE8A | PRKAR2A | RPS6KA3 | WNK1 |
| CCND1 | DUSP11 | ICAM1 | MELK | PDGFRB | PRKCA | RPS6KA4 | YES1 |
| CCT2 | DUSP12 | ICK | MET | PDK1 | PRKCI | RPS6KB1 | ZMPSTE24 |
| CD63 | DVL2 | IGF1R | MIF | PGK1 | PRKCZ | RYK | |
| CD82 | DVL3 | IGF2R | MINPP1 | PHPT1 | PRKDC | SHC1 | |
Related Materials
Response not successful: Received status code 500
Select Citations
If additional help is needed, please consult our technical services group at [email protected].
MISSION is a trademark of Merck KGaA, Darmstadt, Germany and/or its affiliates. Label License.